Search
Search

Rpf1| Gene Symbol: Rpf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100794.1 | NP_001094264.1 | 6 | 704 | ||
| GenBank mRNA | BC086515.1 | - | 6 | 673 | ||
| BC166888.1 | - | 6 | 701 | 6891440 | ||
| BC166920.1 | - | 6 | 701 | 6919256 | ||
| DY564389.1 | - | 6 | 682 | |||
| FQ220191.1 | - | 6 | 683 | |||
| FQ226064.1 | - | 6 | 690 | |||
| FQ227364.1 | - | 6 | 690 | |||
| FQ234387.1 | - | 6 | 690 | |||