Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DLC1 SIRNA Assays ›
Gene Symbol:
Dlc1
Gene Name:
DLC1 Rho GTPase activating protein
Gene Aliases:
Arhgap7, RhoGAP
Chromosome Location:
Chr.16: 59197367 - 59245987 on Build Rnor_6.0
UniGene ID:
Rn.224563
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
RSS367604
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Dlc1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001127446.1
NP_001120918.1
12
3124
GenBank mRNA
D31962.1
-
12
3124
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001194941, NM_001194940, NM_015802, AF178078, AK166175, and others.
Panther Classification:
Molecular Function -
G-protein modulator
protein-binding activity modulator
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
neural tube closure
heart morphogenesis
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
cytoskeleton organization
signal transduction
activation of phospholipase C activity
Rho protein signal transduction
negative regulation of cell proliferation
regulation of cell shape
hindbrain morphogenesis
actin cytoskeleton organization
negative regulation of cell migration
forebrain development
cellular response to insulin stimulus
regulation of actin cytoskeleton organization
negative regulation of Rho protein signal transduction
positive regulation of protein dephosphorylation
focal adhesion assembly
negative regulation of stress fiber assembly
negative regulation of focal adhesion assembly
positive regulation of execution phase of apoptosis
GTPase activator activity
lipid binding
vinculin binding
SH2 domain binding
phospholipase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0