Search
Search

Fxyd6| Gene Symbol: Fxyd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022005.2 | NP_071288.1 | 4 | 374 | ||
| XM_008766171.1 | XP_008764393.1 | 4 | 403 | |||
| GenBank mRNA | AA799380.1 | - | 4 | 324 | ||
| AB030908.1 | - | 4 | 188 | |||
| AF142439.1 | - | 4 | 184 | |||
| BC072528.1 | - | 4 | 180 | 7096832 | ||
| BM385427.1 | - | 6 | 511 | |||
| CB741544.1 | - | 4 | 396 | |||