Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPLP0 SIRNA Assays ›
          Gene Symbol:
          Rplp0
          Gene Name:
          ribosomal protein, large, P0
          Gene Aliases:
          Arbp
          Chromosome Location:
          Chr.12: 46791528 - 46794797 on Build Rnor_6.0
          UniGene ID:
          Rn.1079
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID RSS368056
          Size 20 nmol
          Availability Made To Order
          Catalog # 1330001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rplp0
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_022402.2 NP_071797.1 5 572
          GenBank mRNA BC062028.1 - 4 541 5623881
          DN931898.1 - 4 590
          FQ209603.1 - 5 572
          FQ209620.1 - 4 557
          FQ209649.1 - 5 572
          FQ209757.1 - 5 573
          FQ210253.1 - 5 572
          FQ210412.1 - 5 572
          FQ211152.1 - 5 572
          FQ211263.1 - 5 571
          FQ212554.1 - 5 572
          FQ212743.1 - 5 572
          FQ212875.1 - 5 572
          FQ212927.1 - 5 573
          FQ213085.1 - 5 572
          FQ213427.1 - 5 572
          FQ213510.1 - 5 572
          FQ213712.1 - 5 572
          FQ213847.1 - 5 573
          FQ214087.1 - 5 572
          FQ214354.1 - 5 572
          FQ214515.1 - 5 572
          FQ214553.1 - 5 573
          FQ214850.1 - 5 572
          FQ214997.1 - 5 572
          FQ215007.1 - 5 572
          FQ215548.1 - 5 574
          FQ215662.1 - 5 572
          FQ215682.1 - 5 572
          FQ215870.1 - 5 572
          FQ216120.1 - 5 573
          FQ216296.1 - 5 572
          FQ216389.1 - 5 573
          FQ216406.1 - 5 575
          FQ216495.1 - 5 572
          FQ216618.1 - 5 572
          FQ216655.1 - 5 573
          FQ216733.1 - 5 574
          FQ216774.1 - 5 572
          FQ216813.1 - 5 571
          FQ216851.1 - 5 572
          FQ217406.1 - 5 572
          FQ218561.1 - 5 572
          FQ218832.1 - 5 572
          FQ218943.1 - 5 572
          FQ219025.1 - 5 575
          FQ219184.1 - 5 576
          FQ219389.1 - 5 572
          FQ219457.1 - 5 572
          FQ219678.1 - 5 572
          FQ219815.1 - 5 572
          FQ219817.1 - 5 572
          FQ219837.1 - 5 572
          FQ219855.1 - 5 572
          FQ219923.1 - 5 572
          FQ220008.1 - 5 572
          FQ220051.1 - 5 572
          FQ220077.1 - 5 572
          FQ220115.1 - 5 572
          FQ220181.1 - 5 572
          FQ220201.1 - 5 570
          FQ220213.1 - 5 572
          FQ220247.1 - 5 573
          FQ220282.1 - 5 574
          FQ220284.1 - 5 572
          FQ220302.1 - 5 572
          FQ220338.1 - 5 572
          FQ220413.1 - 5 572
          FQ220420.1 - 5 573
          FQ220433.1 - 5 569
          FQ220444.1 - 5 572
          FQ220445.1 - 5 568
          FQ220449.1 - 5 572
          FQ220500.1 - 5 572
          FQ220514.1 - 5 572
          FQ220527.1 - 5 572
          FQ220533.1 - 5 604
          FQ220542.1 - 5 572
          FQ220545.1 - 5 572
          FQ220634.1 - 5 572
          FQ220654.1 - 5 572
          FQ220663.1 - 5 572
          FQ220684.1 - 5 573
          FQ220690.1 - 5 574
          FQ220769.1 - 5 572
          FQ220787.1 - 5 572
          FQ220801.1 - 5 572
          FQ220833.1 - 5 572
          FQ220838.1 - 5 572
          FQ220888.1 - 5 572
          FQ220932.1 - 5 573
          FQ221146.1 - 5 572
          FQ221380.1 - 5 572
          FQ221533.1 - 5 573
          FQ221556.1 - 5 570
          FQ222181.1 - 5 572
          FQ222327.1 - 5 572
          FQ222772.1 - 5 572
          FQ222906.1 - 5 572
          FQ223121.1 - 5 572
          FQ223487.1 - 5 572
          FQ224547.1 - 5 572
          FQ224834.1 - 5 572
          FQ224846.1 - 5 572
          FQ225049.1 - 5 572
          FQ225170.1 - 5 572
          FQ225608.1 - 5 572
          FQ225654.1 - 5 574
          FQ225860.1 - 5 574
          FQ225886.1 - 5 572
          FQ226306.1 - 5 574
          FQ227381.1 - 5 572
          FQ227719.1 - 5 572
          FQ227837.1 - 5 572
          FQ227937.1 - 5 573
          FQ227941.1 - 5 572
          FQ227942.1 - 5 692
          FQ228548.1 - 5 572
          FQ228995.1 - 5 572
          FQ228996.1 - 5 572
          FQ229139.1 - 5 574
          FQ229159.1 - 5 572
          FQ229164.1 - 5 573
          FQ229208.1 - 5 572
          FQ229241.1 - 5 572
          FQ229244.1 - 5 572
          FQ229248.1 - 3 452
          FQ229272.1 - 5 572
          FQ229273.1 - 5 573
          FQ229303.1 - 5 572
          FQ229337.1 - 5 572
          FQ229352.1 - 5 572
          FQ229365.1 - 5 572
          FQ229397.1 - 5 572
          FQ229408.1 - 5 573
          FQ229433.1 - 5 572
          FQ229448.1 - 5 572
          FQ229521.1 - 5 572
          FQ229547.1 - 5 572
          FQ229554.1 - 5 572
          FQ229566.1 - 5 568
          FQ229610.1 - 5 572
          FQ229626.1 - 5 569
          FQ229726.1 - 5 572
          FQ229740.1 - 5 572
          FQ229742.1 - 5 572
          FQ229776.1 - 5 572
          FQ229808.1 - 5 572
          FQ229815.1 - 5 572
          FQ229822.1 - 5 572
          FQ229830.1 - 5 574
          FQ229883.1 - 5 574
          FQ229901.1 - 5 572
          FQ229929.1 - 5 572
          FQ229932.1 - 3 470
          FQ229952.1 - 5 572
          FQ229974.1 - 5 572
          FQ230005.1 - 5 572
          FQ230055.1 - 5 571
          FQ230127.1 - 5 572
          FQ230267.1 - 5 575
          FQ230943.1 - 5 572
          FQ231445.1 - 5 574
          FQ231525.1 - 5 572
          FQ231591.1 - 5 574
          FQ231710.1 - 5 573
          FQ231733.1 - 5 573
          FQ231897.1 - 5 574
          FQ231967.1 - 5 572
          FQ232029.1 - 5 573
          FQ232154.1 - 5 572
          FQ232165.1 - 5 572
          FQ232310.1 - 5 571
          FQ232319.1 - 5 573
          FQ232697.1 - 5 574
          FQ232767.1 - 5 572
          FQ232951.1 - 5 572
          FQ233152.1 - 5 571
          FQ233377.1 - 5 572
          FQ233473.1 - 5 572
          FQ233732.1 - 5 572
          FQ233737.1 - 5 572
          FQ233808.1 - 5 572
          FQ234195.1 - 5 572
          FQ234341.1 - 5 572
          FQ234613.1 - 5 572
          FQ234733.1 - 5 572
          FQ234735.1 - 5 462
          FQ234863.1 - 5 572
          FQ235063.1 - 5 572
          FQ235079.1 - 5 572
          FQ235122.1 - 5 572
          FQ235259.1 - 5 572
          X15096.1 - 5 572
          Z29530.1 - 4 528

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ribosomal protein translational protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translational elongation
          ribosome biogenesis
          cellular response to interleukin-4
          structural constituent of ribosome
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0