Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPLP0 SIRNA Assays ›
          Gene Symbol:
          Rplp0
          Gene Name:
          ribosomal protein, large, P0
          Gene Aliases:
          Arbp
          Chromosome Location:
          Chr.12: 46791528 - 46794797 on Build Rnor_6.0
          UniGene ID:
          Rn.1079
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID RSS368057
          Size 20 nmol
          Availability Made To Order
          Catalog # 1330001
          Price 458.00
          Your Price
          Online offer:
          458.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rplp0
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_022402.2 NP_071797.1 5 598
          GenBank mRNA BC062028.1 - 4 567 5623881
          BP465362.1 - 3 492
          DN931898.1 - 4 616
          FQ209537.1 - 5 598
          FQ209603.1 - 5 598
          FQ209620.1 - 4 583
          FQ209649.1 - 5 598
          FQ209757.1 - 5 599
          FQ210412.1 - 5 598
          FQ211152.1 - 5 598
          FQ211263.1 - 5 597
          FQ212554.1 - 5 598
          FQ212743.1 - 5 598
          FQ212875.1 - 5 598
          FQ212927.1 - 5 599
          FQ213085.1 - 5 598
          FQ213427.1 - 5 598
          FQ213510.1 - 5 598
          FQ213712.1 - 5 598
          FQ213847.1 - 5 599
          FQ214553.1 - 5 599
          FQ214850.1 - 5 598
          FQ214997.1 - 5 598
          FQ215007.1 - 5 598
          FQ215548.1 - 5 600
          FQ215662.1 - 5 598
          FQ215682.1 - 5 598
          FQ215870.1 - 5 598
          FQ216120.1 - 5 599
          FQ216296.1 - 5 598
          FQ216389.1 - 5 599
          FQ216406.1 - 5 601
          FQ216495.1 - 5 598
          FQ216618.1 - 5 598
          FQ216655.1 - 5 599
          FQ216733.1 - 5 600
          FQ216774.1 - 5 598
          FQ216813.1 - 5 597
          FQ216851.1 - 5 598
          FQ216913.1 - 5 599
          FQ217406.1 - 5 598
          FQ218561.1 - 5 598
          FQ218832.1 - 5 598
          FQ218943.1 - 5 598
          FQ219025.1 - 5 601
          FQ219184.1 - 5 602
          FQ219389.1 - 5 598
          FQ219457.1 - 5 598
          FQ219678.1 - 5 598
          FQ219817.1 - 5 598
          FQ219837.1 - 5 598
          FQ219855.1 - 5 598
          FQ219923.1 - 5 598
          FQ220008.1 - 5 598
          FQ220051.1 - 5 598
          FQ220077.1 - 5 598
          FQ220115.1 - 5 598
          FQ220181.1 - 5 598
          FQ220201.1 - 5 596
          FQ220213.1 - 5 598
          FQ220247.1 - 5 599
          FQ220282.1 - 5 600
          FQ220284.1 - 5 598
          FQ220302.1 - 5 598
          FQ220338.1 - 5 598
          FQ220345.1 - 5 599
          FQ220370.1 - 5 598
          FQ220406.1 - 5 598
          FQ220413.1 - 5 598
          FQ220420.1 - 5 599
          FQ220433.1 - 5 595
          FQ220444.1 - 5 598
          FQ220445.1 - 5 594
          FQ220449.1 - 5 598
          FQ220500.1 - 5 598
          FQ220514.1 - 5 598
          FQ220527.1 - 5 598
          FQ220533.1 - 5 630
          FQ220542.1 - 5 598
          FQ220545.1 - 5 598
          FQ220634.1 - 5 598
          FQ220654.1 - 5 598
          FQ220663.1 - 5 598
          FQ220684.1 - 5 599
          FQ220690.1 - 5 600
          FQ220769.1 - 5 598
          FQ220787.1 - 5 598
          FQ220801.1 - 5 598
          FQ220833.1 - 5 598
          FQ220838.1 - 5 598
          FQ220888.1 - 5 598
          FQ220932.1 - 5 599
          FQ221146.1 - 5 598
          FQ221380.1 - 5 598
          FQ221533.1 - 5 599
          FQ221556.1 - 5 596
          FQ222171.1 - 5 598
          FQ222181.1 - 5 598
          FQ222327.1 - 5 598
          FQ222772.1 - 5 598
          FQ222906.1 - 5 598
          FQ223121.1 - 5 598
          FQ223487.1 - 5 598
          FQ224547.1 - 5 598
          FQ224834.1 - 5 598
          FQ224846.1 - 5 598
          FQ225049.1 - 5 598
          FQ225170.1 - 5 598
          FQ225608.1 - 5 598
          FQ225654.1 - 5 600
          FQ225860.1 - 5 600
          FQ225886.1 - 5 598
          FQ226306.1 - 5 600
          FQ227381.1 - 5 598
          FQ227522.1 - 5 593
          FQ227719.1 - 5 598
          FQ227837.1 - 5 598
          FQ227937.1 - 5 599
          FQ227941.1 - 5 598
          FQ227942.1 - 5 718
          FQ228548.1 - 5 598
          FQ228995.1 - 5 598
          FQ228996.1 - 5 598
          FQ229139.1 - 5 600
          FQ229159.1 - 5 598
          FQ229164.1 - 5 599
          FQ229208.1 - 5 598
          FQ229241.1 - 5 598
          FQ229244.1 - 5 598
          FQ229248.1 - 3 478
          FQ229272.1 - 5 598
          FQ229273.1 - 5 599
          FQ229303.1 - 5 598
          FQ229337.1 - 5 598
          FQ229352.1 - 5 598
          FQ229365.1 - 5 598
          FQ229397.1 - 5 598
          FQ229408.1 - 5 599
          FQ229433.1 - 5 598
          FQ229448.1 - 5 598
          FQ229521.1 - 5 598
          FQ229547.1 - 5 598
          FQ229554.1 - 5 598
          FQ229566.1 - 5 594
          FQ229610.1 - 5 598
          FQ229626.1 - 5 595
          FQ229726.1 - 5 598
          FQ229740.1 - 5 598
          FQ229742.1 - 5 598
          FQ229776.1 - 5 598
          FQ229808.1 - 5 598
          FQ229815.1 - 5 598
          FQ229822.1 - 5 598
          FQ229830.1 - 5 600
          FQ229883.1 - 5 600
          FQ229901.1 - 5 598
          FQ229929.1 - 5 598
          FQ229952.1 - 5 598
          FQ229974.1 - 5 598
          FQ230005.1 - 5 598
          FQ230055.1 - 5 597
          FQ230127.1 - 5 598
          FQ230267.1 - 5 601
          FQ230943.1 - 5 598
          FQ231445.1 - 5 600
          FQ231525.1 - 5 598
          FQ231591.1 - 5 600
          FQ231710.1 - 5 599
          FQ231733.1 - 5 599
          FQ231897.1 - 5 600
          FQ231967.1 - 5 598
          FQ232029.1 - 5 599
          FQ232154.1 - 5 598
          FQ232165.1 - 5 598
          FQ232310.1 - 5 597
          FQ232319.1 - 5 599
          FQ232697.1 - 5 600
          FQ232767.1 - 5 598
          FQ232951.1 - 5 598
          FQ233152.1 - 5 597
          FQ233377.1 - 5 598
          FQ233473.1 - 5 598
          FQ233732.1 - 5 598
          FQ233737.1 - 5 598
          FQ233808.1 - 5 598
          FQ234195.1 - 5 598
          FQ234341.1 - 5 598
          FQ234613.1 - 5 598
          FQ234733.1 - 5 598
          FQ234735.1 - 5 488
          FQ234863.1 - 5 598
          FQ235063.1 - 5 598
          FQ235079.1 - 5 598
          FQ235122.1 - 5 598
          FQ235259.1 - 5 598
          X15096.1 - 5 598
          Z29530.1 - 4 554

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ribosomal protein translational protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translational elongation
          ribosome biogenesis
          cellular response to interleukin-4
          structural constituent of ribosome
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0