Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CYBA SIRNA Assays ›
Gene Symbol:
Cyba
Gene Name:
cytochrome b-245, alpha polypeptide
Gene Aliases:
p22-phox
Chromosome Location:
Chr.19: 55249634 - 55257824 on Build Rnor_6.0
UniGene ID:
Rn.5856
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
RSS372687
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Cyba
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_024160.1
NP_077074.1
2
157
GenBank mRNA
AJ295951.1
-
2
143
U18729.1
-
2
157
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of endothelial cell proliferation
negative regulation of glomerular filtration by angiotensin
superoxide metabolic process
inflammatory response
regulation of blood pressure
smooth muscle hypertrophy
cytochrome complex assembly
positive regulation of cell growth
response to nutrient levels
positive regulation of superoxide anion generation
response to drug
superoxide anion generation
innate immune response
respiratory burst
positive regulation of smooth muscle cell proliferation
hydrogen peroxide biosynthetic process
regulation of release of sequestered calcium ion into cytosol
oxidation-reduction process
positive regulation of mucus secretion
response to interleukin-1
cellular response to amino acid stimulus
cellular response to mechanical stimulus
cellular response to organic substance
cellular response to glucose stimulus
cellular response to tumor necrosis factor
cellular response to organic cyclic compound
cellular response to gamma radiation
reactive oxygen species metabolic process
protein binding
electron carrier activity
superoxide-generating NADPH oxidase activity
SH3 domain binding
heme binding
metal ion binding
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-nbkcc:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0