Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DNMT1 SIRNA Assays ›
Gene Symbol:
Dnmt1
Gene Name:
DNA (cytosine-5-)-methyltransferase 1
Gene Aliases:
-
Chromosome Location:
Chr.8: 21922515 - 21968495 on Build Rnor_6.0
UniGene ID:
Rn.6955
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
RSS373066
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Dnmt1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_053354.3
NP_445806.3
19
1581
GenBank mRNA
AB012214.1
-
19
1565
EV775426.1
-
212
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA metabolism protein
DNA methyltransferase
nucleic acid metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
DNA methylation
transcription, DNA-templated
regulation of transcription, DNA-templated
brain development
aging
response to heat
response to toxic substance
response to organic substance
response to ionizing radiation
maintenance of DNA methylation
response to lead ion
DNA methylation on cytosine within a CG sequence
regulation of gene expression
positive regulation of gene expression
response to activity
gene silencing
chromatin modification
neuron differentiation
response to caffeine
response to nutrient levels
response to estradiol
response to lipopolysaccharide
DNA methylation on cytosine
response to vitamin A
response to testosterone
cellular response to platelet-derived growth factor stimulus
response to antidepressant
regulation of cell proliferation
response to drug
DNA hypermethylation
response to ethanol
negative regulation of transcription, DNA-templated
S-adenosylhomocysteine metabolic process
S-adenosylmethioninamine metabolic process
S-adenosylmethionine metabolic process
positive regulation of histone H3-K4 methylation
negative regulation of histone H3-K9 methylation
cellular response to amino acid stimulus
cellular response to lead ion
cellular response to transforming growth factor beta stimulus
C-5 methylation of cytosine
cellular response to nerve growth factor stimulus
DNA binding
chromatin binding
double-stranded DNA binding
RNA binding
DNA (cytosine-5-)-methyltransferase activity
protein binding
transcription factor binding
methyltransferase activity
zinc ion binding
methyl-CpG binding
S-adenosylmethionine-dependent methyltransferase activity
DNA-methyltransferase activity
protein domain specific binding
estrogen receptor binding
histone deacetylase binding
unmethylated CpG binding
DNA (cytosine-5-)-methyltransferase activity, acting on CpG substrates
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline