Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MKNK1 SIRNA Assays ›
          Gene Symbol:
          MKNK1
          Gene Name:
          MAP kinase interacting serine/threonine kinase 1
          Gene Aliases:
          MNK1
          Chromosome Location:
          Chr.1: 46557407 - 46607140 on Build GRCh38
          UniGene ID:
          Hs.371594
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID VHS40571
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MKNK1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001135553.2 NP_001129025.1 3, 4 370
          NM_003684.5 NP_003675.2 3, 4 370
          NM_198973.3 NP_945324.1 3, 4 328
          NR_024174.2 3, 4 370
          NR_024176.2 4, 5 471
          XM_006711000.1 XP_006711063.1 3, 4 323
          XM_006711001.2 XP_006711064.1 4, 5 573
          XM_006711002.2 XP_006711065.1 3, 4 5798
          XM_006711003.1 XP_006711066.1 4, 5 384
          XM_006711013.1 XP_006711076.1 3, 4 322
          XM_006711014.1 XP_006711077.1 3, 4 313
          XM_006711015.1 XP_006711078.1 4, 5 384
          XM_017002655.1 XP_016858144.1 4, 5 6144
          XM_017002656.1 XP_016858145.1 4, 5 5943
          XM_017002659.1 XP_016858148.1 3, 4 342
          XM_017002660.1 XP_016858149.1 3, 4 5798
          XM_017002661.1 XP_016858150.1 4, 5 563
          XM_017002662.1 XP_016858151.1 4, 5 384
          XM_017002663.1 XP_016858152.1 4, 5 6144
          XM_017002664.1 XP_016858153.1 3, 4 5798
          XM_017002666.1 XP_016858155.1 4, 5 3273
          XM_017002667.1 XP_016858156.1 2, 3 849
          GenBank mRNA AB000409.1 - 3, 4 313
          AB451300.1 - 2, 3 126
          AB451432.1 - 2, 3 126
          AK096423.1 - 4, 5 415
          AK290456.1 - 3, 4 290
          AK298839.1 - 3, 4 344
          AK304005.1 - 3, 4 393
          AY355461.1 - 2, 3 126
          BC002755.2 - 3, 4 290 3629765
          BG722049.1 - 3, 4 333
          BX538193.1 - 2, 3 194
          DA435648.1 - 3, 4 315
          DA502363.1 - 3, 4 343
          DC366212.1 - 3, 4 344
          DC376088.1 - 3, 4 300

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of translational initiation
          protein phosphorylation
          peptidyl-serine phosphorylation
          intracellular signal transduction
          protein autophosphorylation
          protein serine/threonine kinase activity
          calmodulin-dependent protein kinase activity
          protein binding
          calmodulin binding
          ATP binding
          calcium-dependent protein serine/threonine kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-x42j5:80/100.66.73.110:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0