Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SMAD1 SIRNA Assays ›
          Gene Symbol:
          SMAD1
          Gene Name:
          SMAD family member 1
          Gene Aliases:
          BSP-1, BSP1, JV4-1, JV41, MADH1, MADR1
          Chromosome Location:
          Chr.4: 145481306 - 145559176 on Build GRCh38
          UniGene ID:
          Hs.604588
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID VHS41102
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SMAD1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001003688.1 NP_001003688.1 2 607
          NM_005900.2 NP_005891.1 2 783
          XM_005262992.3 XP_005263049.1 2 576
          XM_006714217.2 XP_006714280.1 2 638
          XM_011531961.1 XP_011530263.1 2 598
          XM_011531962.1 XP_011530264.1 2 643
          XM_011531963.1 XP_011530265.1 2 765
          XM_011531964.1 XP_011530266.1 2 795
          GenBank mRNA AK293055.1 - 2 909
          BC001878.1 - 2 708 3546771
          BT007386.1 - 1 367
          U54826.1 - 2 607
          U57456.1 - 1 519
          U59423.1 - 2 799
          U59912.1 - 2 608

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor gene-specific transcriptional regulator

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          ureteric bud development
          mesodermal cell fate commitment
          osteoblast fate commitment
          transcription from RNA polymerase II promoter
          inflammatory response
          signal transduction
          transforming growth factor beta receptor signaling pathway
          SMAD protein complex assembly
          gamete generation
          negative regulation of cell proliferation
          embryonic pattern specification
          positive regulation of gene expression
          BMP signaling pathway
          midbrain development
          hindbrain development
          primary miRNA processing
          homeostatic process
          positive regulation of osteoblast differentiation
          positive regulation of transcription from RNA polymerase II promoter
          cartilage development
          cardiac muscle cell proliferation
          bone development
          SMAD protein signal transduction
          positive regulation of cartilage development
          positive regulation of transcription from RNA polymerase II promoter involved in cellular response to chemical stimulus
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II core promoter sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcription factor activity, sequence-specific DNA binding
          receptor signaling protein activity
          protein binding
          protein kinase binding
          transforming growth factor beta receptor, pathway-specific cytoplasmic mediator activity
          identical protein binding
          protein homodimerization activity
          metal ion binding
          protein heterodimerization activity
          co-SMAD binding
          I-SMAD binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0