Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SNHG3 SIRNA Assays ›
          Gene Symbol:
          Snhg3
          Gene Name:
          small nucleolar RNA host gene 3
          Gene Aliases:
          Rnu17d, U17HG
          Chromosome Location:
          Chr.4: 132351933 - 132353686 on Build GRCm38
          UniGene ID:
          Mm.486919
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n253527

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Snhg3
          Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003270.2
          3 159
          GenBank mRNA AJ006837.1 - LIT1313
          U17HG
          3 106
          AK076188.1 -
          3 107
          BC100513.1 -
          3 152
          CN834544.1 -
          3 162
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BY118837.1 - 3 143 Chr. 4 - 132352035 - 132353668 [-] on Build GRCm38
          GenBankmRNA EL959114.1 - 3 151 Chr. 4 - 132351934 - 132353651 [-] on Build GRCm38
          GenBankmRNA BY118222.1 - 3 142 Chr. 4 - 132352005 - 132353668 [-] on Build GRCm38
          GenBankmRNA CN841655.1 - 3 155 Chr. 4 - 132351934 - 132353679 [-] on Build GRCm38
          GenBankmRNA DV069876.1 - 175 Chr. - - [-] on Build
          GenBankmRNA DV069651.1 - 126 Chr. - - [-] on Build
          GenBankmRNA DV061498.1 - 204 Chr. - - [-] on Build
          GenBankmRNA CA460116.1 - 196 Chr. - - [-] on Build
          GenBankmRNA DV062701.1 - 227 Chr. - - [-] on Build
          GenBankmRNA AI787530.1 - 3 105 Chr. 4 - 132351934 - 132353632 [-] on Build GRCm38
          GenBankmRNA CJ216122.1 - 2 121 Chr. 4 - 132351934 - 132352750 [-] on Build GRCm38
          GenBankmRNA CA456802.1 - 173 Chr. - - [-] on Build
          GenBankmRNA BY144490.1 - 3 103 Chr. 4 - 132351966 - 132353630 [-] on Build GRCm38
          GenBankmRNA BY121084.1 - 3 142 Chr. 4 - 132352035 - 132353668 [-] on Build GRCm38
          GenBankmRNA DV072144.1 - 126 Chr. - - [-] on Build
          GenBankmRNA BE198161.1 - 3 133 Chr. 4 - 132352117 - 132353660 [-] on Build GRCm38
          GenBankmRNA AJ006836.1 - 1 1406 Chr. - - [-] on Build
          GenBankmRNA CA492396.1 - 143 Chr. - - [-] on Build
          GenBankmRNA BY713659.1 - 3 101 Chr. 4 - 132351933 - 132353628 [-] on Build GRCm38
          GenBankmRNA AI788223.1 - 3 105 Chr. 4 - 132351934 - 132353632 [-] on Build GRCm38
          GenBankmRNA BY579581.1 - 2 147 Chr. 4 - 132351934 - 132352776 [-] on Build GRCm38
          GenBankmRNA BY121260.1 - 3 142 Chr. 4 - 132352022 - 132353668 [-] on Build GRCm38
          GenBankmRNA DV041333.1 - 201 Chr. - - [-] on Build
          GenBankmRNA CA786577.1 - 155 Chr. - - [-] on Build

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0