Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IL4 SIRNA Assays ›
Gene Symbol:
Il4
Gene Name:
interleukin 4
Gene Aliases:
BSF-1, Il-4
Chromosome Location:
Chr.11: 53612460 - 53618665 on Build GRCm38
UniGene ID:
Mm.276360
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
n257564
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Il4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NR_027491.1
1, 2
257
GenBank mRNA
AA199406.1
-
1, 2
257
AF359558.1
-
1, 2
196
BX529138.1
-
1, 2
266
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
Transcript Location
GenBankmRNA
AI324371.1
-
1, 2
95
Chr. 11 - 53612469 - 53617176 [-] on Build GRCm38
GenBankmRNA
AA153220.1
-
1, 2
95
Chr. 11 - 53612474 - 53617176 [-] on Build GRCm38
GenBankmRNA
AW988069.1
-
1, 2
254
Chr. 11 - 53612468 - 53617335 [-] on Build GRCm38
GenBankmRNA
AA967539.1
-
257
Chr. - - [-] on Build
GenBankmRNA
BY056727.1
-
1, 2
259
Chr. 11 - 53614000 - 53617338 [-] on Build GRCm38
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
microglial cell activation
positive regulation of protein phosphorylation
innate immune response in mucosa
positive regulation of defense response to virus by host
T-helper 1 cell lineage commitment
positive regulation of immunoglobulin production
negative regulation of acute inflammatory response
negative regulation of chronic inflammatory response
immune response
cholesterol metabolic process
positive regulation of cell proliferation
regulation of proton transport
negative regulation of epithelial cell migration
positive regulation of B cell proliferation
B cell costimulation
positive regulation of interleukin-10 production
positive regulation of interleukin-13 production
T-helper 2 cell cytokine production
positive regulation of T cell proliferation
positive regulation of activated T cell proliferation
B cell activation
positive regulation of tyrosine phosphorylation of Stat5 protein
defense response to protozoan
negative regulation of macrophage activation
positive regulation of mast cell degranulation
negative regulation of nitric oxide biosynthetic process
T-helper 2 cell differentiation
positive regulation of chemokine biosynthetic process
positive regulation of MHC class II biosynthetic process
positive regulation of T cell differentiation
negative regulation of osteoclast differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of isotype switching to IgE isotypes
positive regulation of isotype switching to IgG isotypes
regulation of inflammatory response
positive regulation of peptidyl-tyrosine phosphorylation
regulation of immune response
negative regulation of T cell activation
positive regulation of B cell activation
positive regulation of sequence-specific DNA binding transcription factor activity
positive regulation of mononuclear cell migration
dendritic cell differentiation
extrinsic apoptotic signaling pathway in absence of ligand
positive regulation of myoblast fusion
negative regulation of complement-dependent cytotoxicity
negative regulation of T-helper 17 cell differentiation
negative regulation of endothelial cell apoptotic process
positive regulation of eosinophil chemotaxis
negative regulation of extrinsic apoptotic signaling pathway
cytokine activity
cytokine receptor binding
interleukin-4 receptor binding
growth factor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0