Search
Search

CETN4P| Gene Symbol: CETN4P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024041.1 | 3 | 545 | |||
| GenBank mRNA | AV733354.1 | - | 3 | 545 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ429158.1 | - | 3 | 638 | Chr. 4 - 122730161 - 122732419 [-] on Build GRCh38 | |