Search
Search

LIMS3-LOC440895
LOC100288570| Gene Symbol: LIMS3-LOC440895 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027145.2 | 15 | 2065 | |||
| GenBank mRNA | AK023300.2 | - | 8 | 754 | ||
| AL133596.1 | - | 2 | 1761 | |||
| Gene Symbol: LOC100288570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | CB993644.1 | - | 2 | 184 | ||