Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TMX2 SIRNA Assays ›
          Gene Symbol:
          TMX2
          Gene Name:
          thioredoxin related transmembrane protein 2
          Gene Aliases:
          CGI-31, PDIA12, PIG26, TXNDC14
          Chromosome Location:
          Chr.11: 57712523 - 57740973 on Build GRCh38
          UniGene ID:
          Hs.654965
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n260840

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TMX2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_026593.2 3 404
          GenBank mRNA AK298065.1 - 3 316
          CD690929.1 - 3 348
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BP199522.1 - 3 312 Chr. 11 - 57712615 - 57738717 [+] on Build GRCh38
          GenBankmRNA DC303094.1 - 3 314 Chr. 11 - 57712613 - 57738707 [+] on Build GRCh38
          GenBankmRNA BP366894.1 - 3 229 Chr. 11 - 57712699 - 57739006 [+] on Build GRCh38
          GenBankmRNA BM829431.1 - 3 314 Chr. 11 - 57712613 - 57738766 [+] on Build GRCh38
          GenBankmRNA BG753793.1 - 3 275 Chr. 11 - 57712653 - 57740128 [+] on Build GRCh38
          GenBankmRNA BI523672.1 - 3 316 Chr. 11 - 57712611 - 57739198 [+] on Build GRCh38
          GenBankmRNA BU663672.1 - 3 278 Chr. 11 - 57712649 - 57738067 [+] on Build GRCh38
          GenBankmRNA DC300649.1 - 3 314 Chr. 11 - 57712613 - 57738710 [+] on Build GRCh38
          GenBankmRNA DC312211.1 - 3 315 Chr. 11 - 57712614 - 57738680 [+] on Build GRCh38
          GenBankmRNA CD684754.1 - 3 350 Chr. 11 - 57712600 - 57738714 [+] on Build GRCh38
          GenBankmRNA BI223383.1 - 3 303 Chr. 11 - 57712624 - 57739003 [+] on Build GRCh38
          GenBankmRNA DC397847.1 - 3 314 Chr. 11 - 57712613 - 57738694 [+] on Build GRCh38
          GenBankmRNA BI908111.1 - 3 238 Chr. 11 - 57712690 - 57739192 [+] on Build GRCh38
          GenBankmRNA DC311881.1 - 3 319 Chr. 11 - 57712614 - 57738433 [+] on Build GRCh38
          GenBankmRNA DC302616.1 - 3 314 Chr. 11 - 57712614 - 57738717 [+] on Build GRCh38
          GenBankmRNA BG170274.1 - 206 Chr. - - [-] on Build
          GenBankmRNA DC326958.1 - 3 314 Chr. 11 - 57712613 - 57738700 [+] on Build GRCh38
          GenBankmRNA BP286013.1 - 3 319 Chr. 11 - 57712607 - 57738158 [+] on Build GRCh38
          GenBankmRNA DC358840.1 - 3 316 Chr. 11 - 57712614 - 57738719 [+] on Build GRCh38
          GenBankmRNA DA092970.1 - 3 323 Chr. 11 - 57712604 - 57738061 [+] on Build GRCh38
          GenBankmRNA BI906700.1 - 3 313 Chr. 11 - 57712614 - 57739179 [+] on Build GRCh38
          GenBankmRNA BM837220.1 - 3 322 Chr. 11 - 57712604 - 57738779 [+] on Build GRCh38
          GenBankmRNA BG708358.1 - 3 321 Chr. 11 - 57712608 - 57739260 [+] on Build GRCh38
          GenBankmRNA BG541020.1 - 288 Chr. - - [-] on Build
          GenBankmRNA DB491286.1 - 3 315 Chr. 11 - 57712614 - 57737975 [+] on Build GRCh38
          GenBankmRNA CN363044.1 - 3 328 Chr. 11 - 57712612 - 57739212 [+] on Build GRCh38
          GenBankmRNA DC338451.1 - 3 314 Chr. 11 - 57712613 - 57738699 [+] on Build GRCh38
          GenBankmRNA BX419377.2 - 325 Chr. - - [-] on Build
          GenBankmRNA DC408481.1 - 3 309 Chr. 11 - 57712618 - 57738387 [+] on Build GRCh38
          GenBankmRNA BX419011.2 - 3 307 Chr. 11 - 57712620 - 57740283 [+] on Build GRCh38
          GenBankmRNA BI909873.1 - 3 319 Chr. 11 - 57712608 - 57739181 [+] on Build GRCh38
          GenBankmRNA BM172173.1 - 3 321 Chr. 11 - 57712608 - 57738745 [+] on Build GRCh38

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          cell redox homeostasis
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0