Search
Search

MGAM2| Gene Symbol: MGAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293626.1 | NP_001280555.1 | 34 | 4000 | ||
| XM_011516692.2 | XP_011514994.1 | 27 | 3215 | |||
| XM_011516693.2 | XP_011514995.1 | 13 | 1480 | |||
| XM_011516694.2 | XP_011514996.1 | 4 | 341 | |||