Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MAFG-AS1 SIRNA Assays ›
          Gene Symbol:
          MAFG-AS1
          Gene Name:
          MAFG antisense RNA 1 (head to head)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 81927829 - 81930753 on Build GRCh38
          UniGene ID:
          Hs.336958
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n261869

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MAFG-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_015454.1 2 855
          GenBank mRNA BC038198.1 - 2 283
          BC046361.1 - 2 742
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BG475026.1 - 279 Chr. - - [-] on Build
          GenBankmRNA BE389723.1 - 2 259 Chr. 17 - 81928425 - 81930005 [+] on Build GRCh38
          GenBankmRNA BE391273.1 - 2 259 Chr. 17 - 81928425 - 81930167 [+] on Build GRCh38
          GenBankmRNA BE274387.1 - 2 259 Chr. 17 - 81928425 - 81930237 [+] on Build GRCh38
          GenBankmRNA BF026113.1 - 259 Chr. - - [-] on Build
          GenBankmRNA BF027326.1 - 2 280 Chr. 17 - 81928410 - 81930019 [+] on Build GRCh38
          GenBankmRNA BX332192.2 - 2 539 Chr. 17 - 81928144 - 81930176 [+] on Build GRCh38
          GenBankmRNA BE897907.1 - 2 263 Chr. 17 - 81928421 - 81930463 [+] on Build GRCh38
          GenBankmRNA BE387828.1 - 2 259 Chr. 17 - 81928425 - 81930088 [+] on Build GRCh38
          GenBankmRNA BE384188.1 - 2 259 Chr. 17 - 81928425 - 81930091 [+] on Build GRCh38
          GenBankmRNA BG104375.1 - 2 279 Chr. 17 - 81928406 - 81930190 [+] on Build GRCh38
          GenBankmRNA BE728020.1 - 1 261 Chr. 17 - 81929474 - 81930320 [+] on Build GRCh38
          GenBankmRNA BE276952.1 - 2 259 Chr. 17 - 81928425 - 81930078 [+] on Build GRCh38
          GenBankmRNA BF125587.1 - 259 Chr. - - [-] on Build
          GenBankmRNA BG334057.1 - 2 279 Chr. 17 - 81928406 - 81930167 [+] on Build GRCh38
          GenBankmRNA BE388604.1 - 2 259 Chr. 17 - 81928425 - 81930126 [+] on Build GRCh38
          GenBankmRNA BE388073.1 - 2 260 Chr. 17 - 81928425 - 81929998 [+] on Build GRCh38
          GenBankmRNA BE385607.1 - 2 259 Chr. 17 - 81928425 - 81930085 [+] on Build GRCh38
          GenBankmRNA BG334905.1 - 2 279 Chr. 17 - 81928406 - 81930168 [+] on Build GRCh38
          GenBankmRNA BE385205.1 - 2 259 Chr. 17 - 81928425 - 81930045 [+] on Build GRCh38
          GenBankmRNA BE384468.1 - 2 259 Chr. 17 - 81928425 - 81930090 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0