Search
Search

| Gene Symbol: KRT16P3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029393.1 | 5 | 1588 | |||
| GenBank mRNA | BC110641.1 | - | 5 | 1571 | ||
| Gene Symbol: KRT16P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_073414.1 | 6 | 1288 | |||