Search
Search

| Gene Symbol: LOC642929 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027472.1 | 3, 4 | 555 | |||
| GenBank mRNA | DQ786189.1 | - | 3, 4 | 554 | ||
| Gene Symbol: LOC442028 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037597.1 | 4, 5 | 315 | |||
| GenBank mRNA | AK126106.1 | - | 4, 5 | 315 | ||
| BC042429.1 | - | 4, 5 | 315 | 4822931 | ||