Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PGM5P2 SIRNA Assays ›
          Gene Symbol:
          PGM5P2 FRG1JP
          Gene Name:
          phosphoglucomutase 5 pseudogene 2
          FSHD region gene 1 family member J, pseudogene
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.9: 41007011 - 41074625 on Build GRCh38
          Chromosome Location:
          Chr.9: 63832050 - 63859634 on Build GRCh38
          UniGene ID:
          Hs.571593
          Hs.650100
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID n265977

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PGM5P2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002836.2 9 1515
          GenBank mRNA BC025351.1 - 9 1507
          Gene Symbol: FRG1JP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BM979847.1 - 6 699
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA AL698608.1 - 1 93 Chr. 2 - 38009644 - 38010245 [+] on Build GRCh38
          GenBankmRNA AI002286.1 - 1 170 Chr. 16 - 35230597 - 35231018 [+] on Build GRCh38
          GenBankmRNA DA100091.1 - 1 313 Chr. 15 - 43650728 - 43651271 [-] on Build GRCh38
          GenBankmRNA DA316269.1 - 1 243 Chr. 14 - 54948846 - 54949383 [-] on Build GRCh38
          GenBankmRNA HY295418.1 - 1 306 Chr. 9 - 91230273 - 91230721 [+] on Build GRCh38
          GenBankmRNA AA577726.1 - 1 499 Chr. 2 - 19163135 - 19163662 [+] on Build GRCh38
          GenBankmRNA AL705773.1 - 1 602 Chr. 16 - 47348131 - 47348780 [-] on Build GRCh38
          GenBankmRNA AA774117.1 - 1 113 Chr. 6 - 133146985 - 133147395 [+] on Build GRCh38
          GenBankmRNA AA807733.1 - 1 109 Chr. 3 - 88508011 - 88508227 [+] on Build GRCh38
          GenBankmRNA DA308367.1 - 1 306 Chr. 15 - 42764992 - 42765552 [-] on Build GRCh38
          GenBankmRNA AI380050.1 - 1 309 Chr. 4 - 90228555 - 90228918 [-] on Build GRCh38
          GenBankmRNA BU675894.1 - 6 699 Chr. 9 - 63832050 - 63844382 [+] on Build GRCh38
          GenBankmRNA AV731015.1 - 1 129 Chr. 10 - 115535513 - 115536159 [+] on Build GRCh38
          GenBankmRNA AA523362.1 - 1 122 Chr. 15 - 63621895 - 63622089 [+] on Build GRCh38
          GenBankmRNA DB288875.1 - 1 310 Chr. 16 - 47183619 - 47184084 [-] on Build GRCh38
          GenBankmRNA DB356793.1 - 1 312 Chr. 19 - 12009503 - 12010068 [+] on Build GRCh38
          GenBankmRNA AI804081.1 - 1 309 Chr. 8 - 73056818 - 73057203 [-] on Build GRCh38
          GenBankmRNA AI635977.1 - 1 310 Chr. 15 - 63621705 - 63622135 [+] on Build GRCh38
          GenBankmRNA N70045.1 - 3, 4 311 Chr. 12 - 51328571 - 51345872 [-] on Build GRCh38
          GenBankmRNA AA628668.1 - 1 148 Chr. 8 - 73056791 - 73057042 [-] on Build GRCh38
          GenBankmRNA AL704608.1 - 1 325 Chr. 1 - 94511692 - 94512246 [+] on Build GRCh38
          GenBankmRNA BX954043.1 - 3 328 Chr. 6 - 19496061 - 19766208 [-] on Build GRCh38
          GenBankmRNA AI564481.1 - 1 313 Chr. 14 - 30664365 - 30664843 [-] on Build GRCh38
          GenBankmRNA BX480838.1 - 1 293 Chr. 6 - 56586898 - 56587342 [-] on Build GRCh38
          GenBankmRNA AL580086.3 - 4 696 Chr. 9 - 40993279 - 41014097 [-] on Build GRCh38
          GenBankmRNA BX953697.1 - 1 474 Chr. 3 - 159396872 - 159397356 [+] on Build GRCh38
          GenBankmRNA CB996823.1 - 1 272 Chr. 8 - 98596093 - 98651724 [-] on Build GRCh38
          GenBankmRNA CN269596.1 - 1 650 Chr. 9 - 68279686 - 68280398 [+] on Build GRCh38
          GenBankmRNA AI081365.1 - 1 305 Chr. 9 - 34766420 - 34766828 [+] on Build GRCh38
          GenBankmRNA BE646483.1 - 1 307 Chr. 13 - 41046997 - 41047461 [+] on Build GRCh38
          GenBankmRNA AI366222.1 - 1 111 Chr. 9 - 63844245 - 63859178 [+] on Build GRCh38
          GenBankmRNA BX644260.1 - 1 479 Chr. 4 - 144996619 - 144997196 [-] on Build GRCh38
          GenBankmRNA CN418506.1 - 1 388 Chr. 6 - 75904367 - 75905019 [+] on Build GRCh38
          GenBankmRNA N54364.1 - 1 112 Chr. 9 - 23395573 - 23395804 [+] on Build GRCh38
          GenBankmRNA BG012085.1 - 2 379 Chr. 12 - 39699952 - 39979684 [-] on Build GRCh38
          GenBankmRNA AA281143.1 - 1 309 Chr. 8 - 73056784 - 73057203 [-] on Build GRCh38
          GenBankmRNA DA292048.1 - 1 326 Chr. 15 - 42765003 - 42765572 [-] on Build GRCh38
          GenBankmRNA CB996752.1 - 1 272 Chr. 8 - 98547978 - 98651724 [-] on Build GRCh38
          GenBankmRNA AL704716.1 - 239 Chr. - - [-] on Build
          GenBankmRNA BG202278.1 - 1 66 Chr. 9 - 41131709 - 41151055 [-] on Build GRCh38
          GenBankmRNA CD674449.1 - 1 40 Chr. 15 - 63621975 - 63622512 [+] on Build GRCh38
          GenBankmRNA AU185304.1 - 1 111 Chr. 15 - 63621904 - 63622247 [+] on Build GRCh38
          GenBankmRNA BU676599.1 - 118 Chr. - - [-] on Build

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0