Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PGM5P2 SIRNA Assays ›
          Gene Symbol:
          PGM5P2 HELLPAR LINC00506
          Gene Name:
          phosphoglucomutase 5 pseudogene 2
          HELLP associated long non-coding RNA
          long intergenic non-protein coding RNA 506
          Gene Aliases:
          -
          LINC-HELLP, lncHELLP
          -
          Chromosome Location:
          Chr.9: 41007011 - 41074625 on Build GRCh38
          Chromosome Location:
          -
          Chromosome Location:
          Chr.3: 87089280 - 87157069 on Build GRCh38
          UniGene ID:
          Hs.571593
          Hs.160562
          Hs.570649
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID n265982

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PGM5P2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002836.2 9 1693
          GenBank mRNA BC025351.1 - 9 1685
          Gene Symbol: HELLPAR
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA JX088243.1 - 1 115372
          Gene Symbol: LINC00506
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_047469.1 2 3039
          GenBank mRNA BC030609.1 - 2 3042
          BC050344.1 - 2 3037
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA AJ006345.3 - 2 194460 Chr. 11 - 2444100 - 2849207 [+] on Build GRCh38
          GenBankmRNA AL704608.1 - 1 498 Chr. 1 - 94511692 - 94512246 [+] on Build GRCh38
          GenBankmRNA AF279660.3 - 6 188451 Chr. 13 - 49996975 - 50344526 [+] on Build GRCh38
          GenBankmRNA DA100091.1 - 1 486 Chr. 15 - 43650728 - 43651271 [-] on Build GRCh38
          GenBankmRNA AK022043.1 - 1 571 Chr. 3 - 69343599 - 69346368 [-] on Build GRCh38
          GenBankmRNA DB324275.1 - 1 469 Chr. 6 - 88525924 - 88526486 [+] on Build GRCh38
          GenBankmRNA DA308367.1 - 1 487 Chr. 15 - 42764992 - 42765552 [-] on Build GRCh38
          GenBankmRNA CN418506.1 - 1 561 Chr. 6 - 75904367 - 75905019 [+] on Build GRCh38
          GenBankmRNA AV731015.1 - 1 302 Chr. 10 - 115535513 - 115536159 [+] on Build GRCh38
          GenBankmRNA AA456928.1 - 1 336 Chr. 10 - 59426904 - 59427343 [+] on Build GRCh38
          GenBankmRNA DA292048.1 - 1 507 Chr. 15 - 42765003 - 42765572 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0