Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other JPX SIRNA Assays ›
          Gene Symbol:
          JPX
          Gene Name:
          JPX transcript, XIST activator (non-protein coding)
          Gene Aliases:
          DCBALD06, ENOX, LINC00183, NCRNA00183
          Chromosome Location:
          Chr.X: 73944324 - 74070384 on Build GRCh38
          UniGene ID:
          Hs.648316
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n266321

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 404.00
          Your Price
          Online offer: 391.65
          391.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: JPX
          Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_024582.1
          2 159
          GenBank mRNA AK056172.1 -
          2 141
          AV714079.1 - LIT1011
          Jpx
          144
          BC071776.1 -
          2 155
          BP871721.1 -
          2 108
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BG504771.1 - 2 138 Chr. X - 73944346 - 73947528 [+] on Build GRCh38
          GenBankmRNA DA979750.1 - 2 132 Chr. X - 73944351 - 73947352 [+] on Build GRCh38
          GenBankmRNA BP872745.1 - 2 138 Chr. X - 73944345 - 74004351 [+] on Build GRCh38
          GenBankmRNA BM548256.1 - 191 Chr. - - [-] on Build
          GenBankmRNA BP871836.1 - 2 137 Chr. X - 73944346 - 73947297 [+] on Build GRCh38
          GenBankmRNA CN270680.1 - 2 124 Chr. X - 73944359 - 73999372 [+] on Build GRCh38
          GenBankmRNA AW963219.1 - 131 Chr. - - [-] on Build
          GenBankmRNA AV705096.1 - 2 94 Chr. X - 73944389 - 74004413 [+] on Build GRCh38
          GenBankmRNA BU934969.1 - 2 139 Chr. X - 73944346 - 73999254 [+] on Build GRCh38
          GenBankmRNA AL711317.1 - 2 158 Chr. X - 73944324 - 73999368 [+] on Build GRCh38
          GenBankmRNA BG612854.1 - 2 140 Chr. X - 73944343 - 73999478 [+] on Build GRCh38
          GenBankmRNA HY065667.1 - 2 145 Chr. X - 73944340 - 74010499 [+] on Build GRCh38
          GenBankmRNA CB993733.1 - 168 Chr. - - [-] on Build
          GenBankmRNA BP872499.1 - 2 159 Chr. X - 73944324 - 74004351 [+] on Build GRCh38
          GenBankmRNA BP873595.1 - 2 141 Chr. X - 73944342 - 74004351 [+] on Build GRCh38
          GenBankmRNA BP872256.1 - 2 138 Chr. X - 73944345 - 74004351 [+] on Build GRCh38
          GenBankmRNA CF129475.1 - 2 124 Chr. X - 73944359 - 74004553 [+] on Build GRCh38
          GenBankmRNA F12346.1 - 100 Chr. - - [-] on Build
          GenBankmRNA DA106100.1 - 2 141 Chr. X - 73944342 - 74004417 [+] on Build GRCh38
          GenBankmRNA DA363254.1 - 2 158 Chr. X - 73944326 - 73997077 [+] on Build GRCh38
          GenBankmRNA CN263834.1 - 2 127 Chr. X - 73944356 - 73999314 [+] on Build GRCh38
          GenBankmRNA HY103315.1 - 2 141 Chr. X - 73944342 - 73947268 [+] on Build GRCh38
          GenBankmRNA BM810643.1 - 114 Chr. - - [-] on Build
          GenBankmRNA BU164379.1 - 2 134 Chr. X - 73944349 - 73998832 [+] on Build GRCh38
          GenBankmRNA BP871739.1 - 2 138 Chr. X - 73944345 - 74004345 [+] on Build GRCh38
          GenBankmRNA BP872588.1 - 2 138 Chr. X - 73944345 - 74004345 [+] on Build GRCh38
          GenBankmRNA BP871919.1 - 2 139 Chr. X - 73944344 - 73999613 [+] on Build GRCh38
          GenBankmRNA DA841100.1 - 2 138 Chr. X - 73944345 - 73947361 [+] on Build GRCh38
          GenBankmRNA BG613915.1 - 2 142 Chr. X - 73944342 - 73999860 [+] on Build GRCh38
          GenBankmRNA AL708895.1 - 2 146 Chr. X - 73944338 - 73947333 [+] on Build GRCh38
          GenBankmRNA BP871940.1 - 2 138 Chr. X - 73944345 - 74004351 [+] on Build GRCh38
          GenBankmRNA BU537716.1 - 2 153 Chr. X - 73944333 - 73947334 [+] on Build GRCh38
          GenBankmRNA DA331725.1 - 2 141 Chr. X - 73944342 - 74004540 [+] on Build GRCh38
          GenBankmRNA BP873466.1 - 2 159 Chr. X - 73944324 - 73999111 [+] on Build GRCh38
          GenBankmRNA BP871783.1 - 2 138 Chr. X - 73944345 - 74004351 [+] on Build GRCh38
          GenBankmRNA R19849.1 - 2 122 Chr. X - 73944385 - 74047792 [+] on Build GRCh38
          GenBankmRNA HY172225.1 - 2 142 Chr. X - 73944342 - 73999019 [+] on Build GRCh38
          GenBankmRNA HY088843.1 - 2 145 Chr. X - 73944340 - 74010567 [+] on Build GRCh38
          GenBankmRNA BP872277.1 - 2 159 Chr. X - 73944324 - 73999592 [+] on Build GRCh38
          GenBankmRNA DA103086.1 - 2 140 Chr. X - 73944343 - 74004499 [+] on Build GRCh38
          GenBankmRNA BG435566.1 - 2 156 Chr. X - 73944328 - 74069325 [+] on Build GRCh38
          GenBankmRNA BU589636.1 - 2 140 Chr. X - 73944346 - 73944948 [+] on Build GRCh38
          GenBankmRNA DA427047.1 - 2 157 Chr. X - 73944326 - 73947342 [+] on Build GRCh38
          GenBankmRNA CB991754.1 - 168 Chr. - - [-] on Build
          GenBankmRNA DB240432.1 - 2 137 Chr. X - 73944346 - 73947385 [+] on Build GRCh38
          GenBankmRNA DA881370.1 - 2 136 Chr. X - 73944347 - 73999014 [+] on Build GRCh38
          GenBankmRNA DA679611.1 - 2 141 Chr. X - 73944342 - 73999105 [+] on Build GRCh38
          GenBankmRNA C16093.1 - 1 77 Chr. 3 - 71682388 - 71682656 [-] on Build GRCh38
          GenBankmRNA BP873513.1 - 2 138 Chr. X - 73944345 - 74004351 [+] on Build GRCh38
          GenBankmRNA DA731241.1 - 2 157 Chr. X - 73944326 - 73998845 [+] on Build GRCh38
          GenBankmRNA T32551.1 - 130 Chr. 15 - 42410703 - 42410722 [-] on Build GRCh38
          GenBankmRNA BP872419.1 - 2 159 Chr. X - 73944324 - 74004351 [+] on Build GRCh38
          GenBankmRNA DB250056.1 - 2 137 Chr. X - 73944346 - 73999096 [+] on Build GRCh38
          GenBankmRNA DA700446.1 - 2 141 Chr. X - 73944342 - 73947318 [+] on Build GRCh38
          GenBankmRNA BF686802.1 - 122 Chr. - - [-] on Build
          GenBankmRNA BG427198.1 - 2 142 Chr. X - 73944342 - 73995436 [+] on Build GRCh38
          GenBankmRNA DW450219.1 - 2 154 Chr. X - 73944342 - 73944645 [+] on Build GRCh38
          GenBankmRNA DW466961.1 - 2 156 Chr. X - 73944340 - 73994872 [+] on Build GRCh38
          GenBankmRNA BP871862.1 - 2 143 Chr. X - 73944340 - 73947374 [+] on Build GRCh38
          GenBankmRNA BP873504.1 - 2 135 Chr. X - 73944356 - 74004351 [+] on Build GRCh38
          GenBankmRNA BP872587.1 - 2 139 Chr. X - 73944344 - 74004351 [+] on Build GRCh38
          GenBankmRNA HY081032.1 - 2 144 Chr. X - 73944339 - 73947244 [+] on Build GRCh38
          GenBankmRNA BP873185.1 - 2 139 Chr. X - 73944344 - 74004355 [+] on Build GRCh38
          GenBankmRNA H84526.1 - 119 Chr. - - [-] on Build
          GenBankmRNA BU599790.1 - 2 144 Chr. X - 73944340 - 74004542 [+] on Build GRCh38
          GenBankmRNA BP872964.1 - 2 138 Chr. X - 73944345 - 74004351 [+] on Build GRCh38
          GenBankmRNA BF797301.1 - 2 126 Chr. X - 73944357 - 73999368 [+] on Build GRCh38
          GenBankmRNA R27273.1 - 123 Chr. - - [-] on Build
          GenBankmRNA BP872836.1 - 2 139 Chr. X - 73944344 - 74004351 [+] on Build GRCh38
          GenBankmRNA DA711535.1 - 2 142 Chr. X - 73944342 - 73947373 [+] on Build GRCh38
          GenBankmRNA CB998281.1 - 168 Chr. - - [-] on Build
          GenBankmRNA BP872490.1 - 2 139 Chr. X - 73944344 - 74004345 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0