Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other POU6F1 SIRNA Assays ›
Gene Symbol:
POU6F1
Gene Name:
POU class 6 homeobox 1
Gene Aliases:
BRN5, MPOU, TCFB1
Chromosome Location:
Chr.12: 51186936 - 51218134 on Build GRCh38
UniGene ID:
Hs.555886
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
n266327
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
5,354.00
Your Price
Online offer:
5354.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: POU6F1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NR_026893.1
2, 3
157
XM_017019507.1
XP_016874996.1
2, 3
1249
XM_017019508.1
XP_016874997.1
3, 4
1160
XM_017019509.1
XP_016874998.1
1, 2
169
XM_017019510.1
XP_016874999.1
2, 3
598
XM_017019511.1
XP_016875000.1
2, 3
767
XM_017019512.1
XP_016875001.1
2, 3
123
XM_017019513.1
XP_016875002.1
2, 3
1249
XM_017019514.1
XP_016875003.1
2, 3
1249
XM_017019515.1
XP_016875004.1
2, 3
595
XM_017019516.1
XP_016875005.1
2, 3
1249
XM_017019517.1
XP_016875006.1
2, 3
1249
XM_017019518.1
XP_016875007.1
2, 3
1249
XM_017019519.1
XP_016875008.1
2, 3
1249
XM_017019520.1
XP_016875009.1
1, 2
39
XM_017019521.1
XP_016875010.1
1, 2
39
XM_017019522.1
XP_016875011.1
1, 2
62
XM_017019524.1
XP_016875013.1
1, 2
169
XM_017019525.1
XP_016875014.1
2, 3
157
XM_017019526.1
XP_016875015.1
1, 2
62
XM_017019527.1
XP_016875016.1
1, 2
62
XM_017019528.1
XP_016875017.1
2, 3
595
XM_017019529.1
XP_016875018.1
2, 3
595
XM_017019530.1
XP_016875019.1
1, 2
500
GenBank mRNA
AI417271.1
-
2, 3
219
AK095191.1
-
2, 3
157
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
Transcript Location
GenBankmRNA
DA784067.1
-
1, 2
39
Chr. 12 - 51198548 - 51206817 [-] on Build GRCh38
GenBankmRNA
DA115694.1
-
2, 3
521
Chr. 12 - 51204321 - 51207574 [-] on Build GRCh38
GenBankmRNA
DB244012.1
-
1, 2
62
Chr. 12 - 51197975 - 51217693 [-] on Build GRCh38
GenBankmRNA
AI283705.1
-
2, 3
131
Chr. 12 - 51198640 - 51217667 [-] on Build GRCh38
GenBankmRNA
DA354451.1
-
2, 3
157
Chr. 12 - 51198571 - 51217693 [-] on Build GRCh38
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
brain development
heart development
muscle organ development
RNA polymerase II regulatory region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-zxk44:80/100.66.78.82:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0