Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BDNF-AS SIRNA Assays ›
          Gene Symbol:
          BDNF-AS
          Gene Name:
          BDNF antisense RNA
          Gene Aliases:
          ANTI-BDNF, BDNF, BDNF-AS1, BDNFOS, NCRNA00049
          Chromosome Location:
          Chr.11: 27506852 - 27698171 on Build GRCh38
          UniGene ID:
          Hs.675323
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n266774

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BDNF-AS
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002832.2 5 437
          NR_033312.1 4 368
          NR_033313.1 4 368
          NR_033314.1 5 437
          NR_033315.1 4 368
          GenBank mRNA AY054391.4 - 5 467
          AY054401.3 - 4 368
          AY054402.3 - 4 368
          AY054403.4 - 5 467
          AY054404.4 - 4 368
          AY054405.4 - 5 467
          AY513486.3 - 4 368
          EF689023.1 - 4 373
          EF689024.1 - 3 276
          EF689025.1 - 3 276
          EF689026.1 - 3 276
          EF689027.1 - 3 276
          EF689028.1 - 3 276
          EF689029.1 - 3 276
          EF689030.1 - 5 442
          EF689031.1 - 5 442
          EF689032.1 - 5 442
          EF689033.1 - 5 442
          EF689034.1 - 5 442
          EF689035.1 - 5 442
          EF689036.1 - 4 373
          EF689037.1 - 4 373
          EF689038.1 - 4 373
          EF689039.1 - 4 373
          EF689040.1 - 4 373
          EF689041.1 - 4 373
          EF689042.1 - 5 442
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA210918.1 - 4 373 Chr. 11 - 27506847 - 27659198 [+] on Build GRCh38
          GenBankmRNA HY129944.1 - 3 275 Chr. 11 - 27506849 - 27658364 [+] on Build GRCh38
          GenBankmRNA DB102543.1 - 5 445 Chr. 11 - 27506844 - 27658410 [+] on Build GRCh38
          GenBankmRNA BI754738.1 - 4 346 Chr. 11 - 27506872 - 27697569 [+] on Build GRCh38
          GenBankmRNA BG707369.1 - 5 466 Chr. 11 - 27506852 - 27675412 [+] on Build GRCh38
          GenBankmRNA DA347710.1 - 5 440 Chr. 11 - 27506852 - 27658439 [+] on Build GRCh38
          GenBankmRNA BG707452.1 - 5 466 Chr. 11 - 27506852 - 27675373 [+] on Build GRCh38
          GenBankmRNA BI821839.1 - 4 347 Chr. 11 - 27506872 - 27675955 [+] on Build GRCh38
          GenBankmRNA DA352679.1 - 4 354 Chr. 11 - 27506838 - 27659200 [+] on Build GRCh38
          GenBankmRNA DA129270.1 - 4 371 Chr. 11 - 27506849 - 27659190 [+] on Build GRCh38
          GenBankmRNA DA422461.1 - 6 542 Chr. 11 - 27506849 - 27658333 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0