Search
Search

NCR3LG1| Gene Symbol: NCR3LG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001202439.2 | NP_001189368.1 | 5 | 2852 | ||
| XM_011520072.2 | XP_011518374.1 | 5 | 4806 | |||
| XM_011520073.2 | XP_011518375.1 | 5 | 4552 | |||
| XM_011520074.2 | XP_011518376.1 | 5 | 3169 | |||
| XM_011520075.2 | XP_011518377.1 | 5 | 4458 | |||
| GenBank mRNA | BX537685.1 | - | 5 | 2844 | ||
| CR749521.1 | - | 5 | 2852 | |||