Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RAMP2-AS1 SIRNA Assays ›
          Gene Symbol:
          RAMP2-AS1
          Gene Name:
          RAMP2 antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 42753914 - 42761257 on Build GRCh38
          UniGene ID:
          Hs.655265
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n268801

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RAMP2-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_024461.1 2 646
          NR_024462.1 3 525
          GenBank mRNA AK057121.1 - 1 674
          BC001607.1 - 2 492
          BC018638.2 - 3 525
          BC033118.1 - 3 525
          BC034924.1 - 2 400
          BC041021.1 - 2 493
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BX283213.1 - 2 493 Chr. 17 - 42759853 - 42761104 [-] on Build GRCh38
          GenBankmRNA BE393341.1 - 3 506 Chr. 17 - 42759738 - 42761238 [-] on Build GRCh38
          GenBankmRNA BE395433.1 - 2 383 Chr. 17 - 42759787 - 42760992 [-] on Build GRCh38
          GENCODE ENST00000591082.5 - 2 508 Chr. 17 - 42756199 - 42761119 [-] on Build GRCh38
          GENCODE ENST00000592195.1 - 2 400 Chr. 17 - 42758295 - 42761011 [-] on Build GRCh38
          GenBankmRNA BE393900.1 - 3 516 Chr. 17 - 42759854 - 42761237 [-] on Build GRCh38
          GenBankmRNA AL550086.3 - 2 696 Chr. 17 - 42755034 - 42761263 [-] on Build GRCh38
          GenBankmRNA BE277594.1 - 2 518 Chr. 17 - 42756210 - 42761100 [-] on Build GRCh38
          GenBankmRNA DA587469.1 - 2 466 Chr. 17 - 42759787 - 42761077 [-] on Build GRCh38
          GenBankmRNA BE389712.1 - 2 460 Chr. 17 - 42759738 - 42761073 [-] on Build GRCh38
          GenBankmRNA DA001969.1 - 2 474 Chr. 17 - 42760426 - 42761085 [-] on Build GRCh38
          GENCODE ENST00000592670.5 - 2 492 Chr. 17 - 42753914 - 42761103 [-] on Build GRCh38
          GenBankmRNA BE277411.1 - 2 462 Chr. 17 - 42756298 - 42761073 [-] on Build GRCh38
          GENCODE ENST00000587694.5 - 1 674 Chr. 17 - 42753931 - 42761117 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0