Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ANKRD36BP2 SIRNA Assays ›
          Gene Symbol:
          ANKRD36BP2
          Gene Name:
          ankyrin repeat domain 36B pseudogene 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 88765902 - 88806596 on Build GRCh38
          UniGene ID:
          Hs.105323
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n269268

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ANKRD36BP2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_015424.1 1 219
          GenBank mRNA AL137717.1 - 1 229
          BC031698.1 - 1 219
          BX648045.1 - 1 253
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BE296318.1 - 1 231 Chr. 2 - 88765890 - 88784861 [+] on Build GRCh38
          GenBankmRNA BM562402.1 - 292 Chr. - - [-] on Build
          GenBankmRNA DB142242.1 - 1 222 Chr. 2 - 88765899 - 88778321 [+] on Build GRCh38
          GenBankmRNA BX282360.1 - 1 196 Chr. 2 - 88765925 - 88782919 [+] on Build GRCh38
          GenBankmRNA DB453579.1 - 1 287 Chr. 2 - 88765834 - 88776566 [+] on Build GRCh38
          GenBankmRNA BI909459.1 - 1 195 Chr. 2 - 88765925 - 88786653 [+] on Build GRCh38
          GenBankmRNA DB052423.1 - 1 215 Chr. 2 - 88765906 - 88784613 [+] on Build GRCh38
          GenBankmRNA DB040773.1 - 1 240 Chr. 2 - 88765881 - 88778305 [+] on Build GRCh38
          GenBankmRNA DB102461.1 - 1 315 Chr. 2 - 88765807 - 88776573 [+] on Build GRCh38
          GenBankmRNA BE270022.1 - 1 231 Chr. 2 - 88765890 - 88784771 [+] on Build GRCh38
          GenBankmRNA DA956346.1 - 1 220 Chr. 2 - 88765904 - 88778336 [+] on Build GRCh38
          GenBankmRNA BI826939.1 - 1 219 Chr. 2 - 88765902 - 88786637 [+] on Build GRCh38
          GenBankmRNA DA923544.1 - 1 217 Chr. 2 - 88765907 - 88784742 [+] on Build GRCh38
          GenBankmRNA DA965554.1 - 1 225 Chr. 2 - 88765897 - 88784616 [+] on Build GRCh38
          GenBankmRNA DB056329.1 - 1 218 Chr. 2 - 88765907 - 88784627 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0