Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RNF216P1 SIRNA Assays ›
          Gene Symbol:
          RNF216P1 RNF216
          Gene Name:
          ring finger protein 216 pseudogene 1
          ring finger protein 216
          Gene Aliases:
          RNF216L, hCG2040556
          CAHH, TRIAD3, U7I1, UBCE7IP1, ZIN
          Chromosome Location:
          Chr.7: 4973985 - 4998169 on Build GRCh38
          Chromosome Location:
          Chr.7: 5620041 - 5781730 on Build GRCh38
          UniGene ID:
          Hs.520636
          Hs.487458
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID n269565

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RNF216P1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_015449.1 1 125
          NR_023384.1 1 125
          NR_023385.1 1 125
          GenBank mRNA AK303831.1 - 1 28
          AK308526.1 - 1 60
          AK309798.1 - 1 66
          AK311517.1 - 1 58
          BC068459.1 - 1 94
          BX538059.1 - 1 52
          DC357738.1 - 1 125
          Gene Symbol: RNF216
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AY062174.1 - 1 37
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BU187037.1 - 1 37 Chr. 7 - 4974073 - 4996911 [+] on Build GRCh38
          GenBankmRNA AL520766.3 - 1 60 Chr. 7 - 4974050 - 4996915 [+] on Build GRCh38
          GenBankmRNA DC413548.1 - 1 28 Chr. 7 - 4974082 - 4996763 [+] on Build GRCh38
          GenBankmRNA BQ926184.1 - 1 38 Chr. 7 - 4974072 - 4996922 [+] on Build GRCh38
          GenBankmRNA CN350417.1 - 1 42 Chr. 7 - 4974068 - 4995525 [+] on Build GRCh38
          GenBankmRNA DR159194.1 - 1 50 Chr. 7 - 4974072 - 4996813 [+] on Build GRCh38
          GenBankmRNA BF338268.1 - 1 56 Chr. 7 - 4974053 - 4996854 [+] on Build GRCh38
          GenBankmRNA DC412017.1 - 1 54 Chr. 7 - 4974056 - 4996725 [+] on Build GRCh38
          GenBankmRNA AL521852.3 - 1 73 Chr. 7 - 4974037 - 4996882 [+] on Build GRCh38
          GenBankmRNA DA730719.1 - 1 79 Chr. 7 - 4974031 - 4995585 [+] on Build GRCh38
          GenBankmRNA BM805546.1 - 1 55 Chr. 7 - 4974067 - 4996987 [+] on Build GRCh38
          GenBankmRNA BI562557.1 - 1 99 Chr. 7 - 4974016 - 4995522 [+] on Build GRCh38
          GenBankmRNA DN998801.1 - 1 51 Chr. 7 - 4974059 - 4996695 [+] on Build GRCh38
          GenBankmRNA DC301966.1 - 1 60 Chr. 7 - 4974050 - 4996693 [+] on Build GRCh38
          GenBankmRNA DA728691.1 - 1 53 Chr. 7 - 4974058 - 4996805 [+] on Build GRCh38
          GenBankmRNA DC383443.1 - 1 30 Chr. 7 - 4974080 - 4996659 [+] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase protein modifying enzyme

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0