Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RNF216P1 SIRNA Assays ›
          Gene Symbol:
          RNF216P1 RNF216
          Gene Name:
          ring finger protein 216 pseudogene 1
          ring finger protein 216
          Gene Aliases:
          RNF216L, hCG2040556
          CAHH, TRIAD3, U7I1, UBCE7IP1, ZIN
          Chromosome Location:
          Chr.7: 4973985 - 4998169 on Build GRCh38
          Chromosome Location:
          Chr.7: 5620041 - 5781730 on Build GRCh38
          UniGene ID:
          Hs.520636
          Hs.487458
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID n269566

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RNF216P1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_015449.1 4 460
          NR_023384.1 4 460
          NR_023385.1 4 460
          GenBank mRNA AK308526.1 - 4 395
          AK309798.1 - 4 401
          AK311517.1 - 4 393
          BC026736.1 - 4 339
          BC068459.1 - 4 429
          BX538059.1 - 4 387
          DC357738.1 - 4 460
          Gene Symbol: RNF216
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AY062174.1 - 4 372
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BU187037.1 - 4 372 Chr. 7 - 4974073 - 4996911 [+] on Build GRCh38
          GenBankmRNA AL520766.3 - 4 395 Chr. 7 - 4974050 - 4996915 [+] on Build GRCh38
          GenBankmRNA BQ926184.1 - 4 373 Chr. 7 - 4974072 - 4996922 [+] on Build GRCh38
          GenBankmRNA BI916700.1 - 3 273 Chr. 7 - 4976312 - 4996882 [+] on Build GRCh38
          GenBankmRNA CN350417.1 - 4 377 Chr. 7 - 4974068 - 4995525 [+] on Build GRCh38
          GenBankmRNA DR159194.1 - 4 385 Chr. 7 - 4974072 - 4996813 [+] on Build GRCh38
          GenBankmRNA BF338268.1 - 4 389 Chr. 7 - 4974053 - 4996854 [+] on Build GRCh38
          GenBankmRNA DC340388.1 - 1 238 Chr. 7 - 4984716 - 4996774 [+] on Build GRCh38
          GenBankmRNA AL521852.3 - 4 408 Chr. 7 - 4974037 - 4996882 [+] on Build GRCh38
          GenBankmRNA DA730719.1 - 4 414 Chr. 7 - 4974031 - 4995585 [+] on Build GRCh38
          GenBankmRNA AA430231.1 - 4 326 Chr. 7 - 4974124 - 4996659 [+] on Build GRCh38
          GenBankmRNA BQ434583.1 - 4 341 Chr. 7 - 4974115 - 4996822 [+] on Build GRCh38
          GenBankmRNA BQ881985.1 - 81 Chr. - - [-] on Build
          GenBankmRNA BI562557.1 - 4 434 Chr. 7 - 4974016 - 4995522 [+] on Build GRCh38
          GenBankmRNA BI871291.1 - 2 124 Chr. 7 - 4983695 - 4997114 [+] on Build GRCh38
          GenBankmRNA BE303029.1 - 2 96 Chr. 7 - 4983722 - 4996754 [+] on Build GRCh38
          GenBankmRNA DN998801.1 - 4 386 Chr. 7 - 4974059 - 4996695 [+] on Build GRCh38
          GenBankmRNA DC301966.1 - 4 396 Chr. 7 - 4974050 - 4996693 [+] on Build GRCh38
          GenBankmRNA DA728691.1 - 4 388 Chr. 7 - 4974058 - 4996805 [+] on Build GRCh38
          GenBankmRNA DC383443.1 - 4 365 Chr. 7 - 4974080 - 4996659 [+] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase protein modifying enzyme

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0