Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FTX SIRNA Assays ›
          Gene Symbol:
          FTX
          Gene Name:
          FTX transcript, XIST regulator (non-protein coding)
          Gene Aliases:
          LINC00182, MIR374AHG, NCRNA00182
          Chromosome Location:
          Chr.X: 74028136 - 74293574 on Build GRCh38
          UniGene ID:
          Hs.349570
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n270178

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FTX
          Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_028379.1
          3 431
          GenBank mRNA AK057701.1 - LIT1007
          Ftx
          3 382
          AK311345.1 -
          3 431
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DC390943.1 - 3 400 Chr. X - 74280427 - 74293543 [-] on Build GRCh38
          GenBankmRNA AA775951.1 - 3 284 Chr. X - 74280474 - 74293427 [-] on Build GRCh38
          GenBankmRNA HY101032.1 - 3 340 Chr. X - 74280456 - 74292556 [-] on Build GRCh38
          GenBankmRNA DA932670.1 - 3 403 Chr. X - 74280432 - 74292362 [-] on Build GRCh38
          GenBankmRNA DA530693.1 - 3 365 Chr. X - 74280345 - 74292583 [-] on Build GRCh38
          GenBankmRNA DB136402.1 - 3 404 Chr. X - 74280444 - 74292363 [-] on Build GRCh38
          GenBankmRNA DA990651.1 - 3 416 Chr. X - 74280449 - 74292633 [-] on Build GRCh38
          GenBankmRNA HY029598.1 - 3 421 Chr. X - 74280983 - 74292376 [-] on Build GRCh38
          GenBankmRNA DC338971.1 - 3 395 Chr. X - 74280407 - 74293536 [-] on Build GRCh38
          GenBankmRNA DC366327.1 - 3 387 Chr. X - 74280401 - 74293530 [-] on Build GRCh38
          GenBankmRNA DC390874.1 - 3 401 Chr. X - 74280445 - 74293539 [-] on Build GRCh38
          GenBankmRNA DB200442.1 - 3 391 Chr. X - 74280402 - 74292351 [-] on Build GRCh38
          GENCODE ENST00000429124.5 - 3 382 Chr. X - 74278374 - 74292600 [-] on Build GRCh38
          GenBankmRNA DC414119.1 - 3 387 Chr. X - 74274374 - 74293530 [-] on Build GRCh38
          GenBankmRNA DC304376.1 - 3 367 Chr. X - 74280385 - 74293509 [-] on Build GRCh38
          GenBankmRNA DA032603.1 - 3 311 Chr. X - 74274356 - 74292529 [-] on Build GRCh38
          GenBankmRNA BI834634.1 - 3 365 Chr. X - 74280420 - 74292325 [-] on Build GRCh38
          GenBankmRNA AV754330.1 - 3 289 Chr. X - 74280301 - 74292508 [-] on Build GRCh38
          GenBankmRNA DB247233.1 - 3 382 Chr. X - 74280215 - 74292600 [-] on Build GRCh38
          GenBankmRNA HY075574.1 - 3 324 Chr. X - 74280447 - 74292284 [-] on Build GRCh38
          GenBankmRNA DA182706.1 - 3 487 Chr. X - 74280930 - 74292036 [-] on Build GRCh38
          GenBankmRNA DC389436.1 - 3 381 Chr. X - 74280380 - 74293524 [-] on Build GRCh38
          GenBankmRNA DA857504.1 - 3 393 Chr. X - 74280405 - 74292353 [-] on Build GRCh38
          GenBankmRNA HY129851.1 - 3 430 Chr. X - 74280967 - 74293573 [-] on Build GRCh38
          GenBankmRNA HY132649.1 - 3 396 Chr. X - 74280930 - 74292355 [-] on Build GRCh38
          GenBankmRNA DA685273.1 - 3 367 Chr. X - 74280411 - 74292585 [-] on Build GRCh38
          GenBankmRNA CB990632.1 - 418 Chr. - - [-] on Build
          GenBankmRNA CN412172.1 - 3 329 Chr. X - 74069358 - 74293472 [-] on Build GRCh38
          GenBankmRNA DA246751.1 - 3 410 Chr. X - 74280411 - 74292628 [-] on Build GRCh38
          GenBankmRNA DC353348.1 - 3 387 Chr. X - 74280461 - 74293530 [-] on Build GRCh38
          GenBankmRNA DA755860.1 - 3 395 Chr. X - 74280408 - 74292355 [-] on Build GRCh38
          GenBankmRNA DB113703.1 - 3 384 Chr. X - 74280400 - 74292343 [-] on Build GRCh38
          GenBankmRNA DB100925.1 - 3 311 Chr. X - 74280126 - 74292529 [-] on Build GRCh38
          GenBankmRNA BG718066.1 - 3 390 Chr. X - 74247864 - 74293527 [-] on Build GRCh38
          GenBankmRNA DC410386.1 - 3 396 Chr. X - 74280431 - 74293539 [-] on Build GRCh38
          GenBankmRNA DC408596.1 - 3 408 Chr. X - 74280944 - 74293550 [-] on Build GRCh38
          GenBankmRNA DB015792.1 - 3 378 Chr. X - 74280396 - 74292596 [-] on Build GRCh38
          GenBankmRNA BM546361.1 - 3 240 Chr. X - 73964008 - 74292342 [-] on Build GRCh38
          GenBankmRNA DB195000.1 - 3 325 Chr. X - 74280390 - 74292542 [-] on Build GRCh38
          GenBankmRNA HY075234.1 - 3 324 Chr. X - 74280433 - 74292284 [-] on Build GRCh38
          GenBankmRNA AW629991.1 - 3 240 Chr. X - 74280461 - 74291789 [-] on Build GRCh38
          GenBankmRNA DC366395.1 - 3 387 Chr. X - 74280934 - 74293530 [-] on Build GRCh38
          GenBankmRNA DC353548.1 - 3 431 Chr. X - 74280930 - 74293574 [-] on Build GRCh38
          GENCODE ENST00000602812.5 - 3 431 Chr. X - 74028136 - 74293574 [-] on Build GRCh38
          GenBankmRNA HY158243.1 - 3 381 Chr. X - 74280930 - 74293524 [-] on Build GRCh38
          GenBankmRNA DA397381.1 - 3 376 Chr. X - 74280393 - 74292594 [-] on Build GRCh38
          GenBankmRNA HY132287.1 - 3 388 Chr. X - 74280930 - 74293530 [-] on Build GRCh38
          GENCODE ENST00000418855.5 - 3 391 Chr. X - 74280401 - 74292351 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-48z6k:80/100.66.78.162:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0