Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NUTM2A-AS1 SIRNA Assays ›
          Gene Symbol:
          NUTM2A-AS1 NUTM2B-AS1
          Gene Name:
          NUTM2A antisense RNA 1
          NUTM2B antisense RNA 1
          Gene Aliases:
          FAM22A-AS1
          -
          Chromosome Location:
          Chr.10: 87238667 - 87342558 on Build GRCh38
          Chromosome Location:
          Chr.10: 79691500 - 79826594 on Build GRCh38
          UniGene ID:
          Hs.440648
          Hs.440648
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID n270293

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NUTM2A-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_024397.1 3, 4 464
          GenBank mRNA AK309777.1 - 3, 4 283
          AK311174.1 - 3, 4 303
          DB257428.1 - 3, 4 465
          Gene Symbol: NUTM2B-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_120611.1 5, 6 702
          NR_120613.1 5, 6 702
          GenBank mRNA CN279211.1 - 5, 6 562
          CN279213.1 - 5, 6 546
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA040049.1 - 3, 4 283 Chr. 10 - 87245755 - 87342377 [-] on Build GRCh38
          GenBankmRNA DA737563.1 - 3, 4 414 Chr. 10 - 87245656 - 87342506 [-] on Build GRCh38
          GenBankmRNA BI767196.1 - 272 Chr. - - [-] on Build
          GenBankmRNA DB456680.1 - 3, 4 391 Chr. 10 - 87246163 - 87342485 [-] on Build GRCh38
          GenBankmRNA DA796822.1 - 3, 4 448 Chr. 10 - 87288387 - 87342541 [-] on Build GRCh38
          GenBankmRNA HY151352.1 - 3, 4 384 Chr. 10 - 87208482 - 87342477 [-] on Build GRCh38
          GenBankmRNA DA829196.1 - 3, 4 261 Chr. 10 - 87287993 - 87342355 [-] on Build GRCh38
          GenBankmRNA DA571282.1 - 3, 4 306 Chr. 10 - 87288087 - 87342400 [-] on Build GRCh38
          GenBankmRNA DA258253.1 - 3, 4 283 Chr. 10 - 87216011 - 87342377 [-] on Build GRCh38
          GenBankmRNA BG111204.1 - 5, 6 363 Chr. 10 - 79691671 - 79826255 [-] on Build GRCh38
          GenBankmRNA DA259512.1 - 3, 4 289 Chr. 10 - 87202973 - 87342383 [-] on Build GRCh38
          GenBankmRNA DA551755.1 - 3, 4 249 Chr. 10 - 87245749 - 87342343 [-] on Build GRCh38
          GenBankmRNA CD250242.1 - 3, 4 471 Chr. 10 - 87288178 - 87342541 [-] on Build GRCh38
          GenBankmRNA HY154971.1 - 3, 4 392 Chr. 10 - 87216096 - 87342485 [-] on Build GRCh38
          GenBankmRNA BM559658.1 - 380 Chr. - - [-] on Build
          GenBankmRNA HY062022.1 - 3, 4 340 Chr. 10 - 87288324 - 87342412 [-] on Build GRCh38
          GenBankmRNA DB263965.1 - 3, 4 276 Chr. 10 - 87206378 - 87342370 [-] on Build GRCh38
          GenBankmRNA DB483325.1 - 3, 4 362 Chr. 10 - 87246167 - 87342456 [-] on Build GRCh38
          GenBankmRNA HY055033.1 - 3, 4 362 Chr. 10 - 87288436 - 87342455 [-] on Build GRCh38
          GenBankmRNA DA009741.1 - 3, 4 281 Chr. 10 - 87245758 - 87342375 [-] on Build GRCh38
          GenBankmRNA AV746236.1 - 3, 4 206 Chr. 10 - 79766294 - 79808834 [-] on Build GRCh38
          GenBankmRNA DA775058.1 - 3, 4 284 Chr. 10 - 87215939 - 87342377 [-] on Build GRCh38
          GenBankmRNA HY087386.1 - 3, 4 340 Chr. 10 - 87288299 - 87342412 [-] on Build GRCh38
          GenBankmRNA HY157097.1 - 3, 4 254 Chr. 10 - 87216111 - 87342347 [-] on Build GRCh38
          GenBankmRNA DA200463.1 - 3, 4 303 Chr. 10 - 87206423 - 87342397 [-] on Build GRCh38
          GenBankmRNA HY150000.1 - 3, 4 282 Chr. 10 - 87216123 - 87342375 [-] on Build GRCh38
          GenBankmRNA T58826.1 - 199 Chr. - - [-] on Build
          GenBankmRNA HY010169.1 - 3, 4 362 Chr. 10 - 87238758 - 87342455 [-] on Build GRCh38
          GenBankmRNA HY124799.1 - 3, 4 254 Chr. 10 - 87216064 - 87342347 [-] on Build GRCh38
          GenBankmRNA BG748770.1 - 4, 5 540 Chr. 10 - 79766290 - 79826477 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0