Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NUTM2A-AS1 SIRNA Assays ›
          Gene Symbol:
          NUTM2A-AS1 NUTM2B-AS1
          Gene Name:
          NUTM2A antisense RNA 1
          NUTM2B antisense RNA 1
          Gene Aliases:
          FAM22A-AS1
          -
          Chromosome Location:
          Chr.10: 87238667 - 87342558 on Build GRCh38
          Chromosome Location:
          Chr.10: 79691500 - 79826594 on Build GRCh38
          UniGene ID:
          Hs.440648
          Hs.440648
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID n270294

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NUTM2A-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_024397.1 1, 2 325
          GenBank mRNA AK309777.1 - 1, 2 144
          AK311174.1 - 1, 2 164
          BC047063.1 - 1, 2 252
          DB257428.1 - 1, 2 326
          Gene Symbol: NUTM2B-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_120611.1 1, 2 405
          NR_120612.1 1, 2 405
          NR_120613.1 1, 2 405
          GenBank mRNA BQ425131.1 - 1, 2 425
          CB163469.1 - 1, 2 222
          CN279211.1 - 1, 2 265
          CN279213.1 - 1, 2 249
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA737563.1 - 1, 2 275 Chr. 10 - 87245656 - 87342506 [-] on Build GRCh38
          GenBankmRNA DA040049.1 - 1, 2 144 Chr. 10 - 87245755 - 87342377 [-] on Build GRCh38
          GenBankmRNA BI767196.1 - 133 Chr. - - [-] on Build
          GenBankmRNA DA796822.1 - 1, 2 309 Chr. 10 - 87288387 - 87342541 [-] on Build GRCh38
          GenBankmRNA AL520958.3 - 1, 2 48 Chr. 10 - 79797377 - 79826237 [-] on Build GRCh38
          GenBankmRNA DA691562.1 - 1, 2 288 Chr. 10 - 79804494 - 79826468 [-] on Build GRCh38
          GenBankmRNA BG111204.1 - 1, 2 66 Chr. 10 - 79691671 - 79826255 [-] on Build GRCh38
          GenBankmRNA DA010830.1 - 2, 3 339 Chr. 10 - 79806056 - 79826563 [-] on Build GRCh38
          GenBankmRNA CD250242.1 - 1, 2 332 Chr. 10 - 87288178 - 87342541 [-] on Build GRCh38
          GenBankmRNA BG166579.1 - 1, 2 304 Chr. 10 - 79766137 - 79826484 [-] on Build GRCh38
          GenBankmRNA HY154971.1 - 1, 2 253 Chr. 10 - 87216096 - 87342485 [-] on Build GRCh38
          GenBankmRNA CN274008.1 - 1, 2 137 Chr. 10 - 79802853 - 79826326 [-] on Build GRCh38
          GenBankmRNA BM559658.1 - 241 Chr. - - [-] on Build
          GenBankmRNA HY062022.1 - 1, 2 201 Chr. 10 - 87288324 - 87342412 [-] on Build GRCh38
          GenBankmRNA DB483325.1 - 1, 2 223 Chr. 10 - 87246167 - 87342456 [-] on Build GRCh38
          GenBankmRNA BG437437.1 - 373 Chr. - - [-] on Build
          GenBankmRNA HY087386.1 - 1, 2 201 Chr. 10 - 87288299 - 87342412 [-] on Build GRCh38
          GenBankmRNA DA200463.1 - 1, 2 164 Chr. 10 - 87206423 - 87342397 [-] on Build GRCh38
          GenBankmRNA CN279215.1 - 1, 2 289 Chr. 10 - 79804484 - 79826470 [-] on Build GRCh38
          GenBankmRNA AW612231.1 - 1, 2 203 Chr. 10 - 79804492 - 79826392 [-] on Build GRCh38
          GenBankmRNA T58826.1 - 60 Chr. - - [-] on Build
          GenBankmRNA HY150000.1 - 1, 2 143 Chr. 10 - 87216123 - 87342375 [-] on Build GRCh38
          GenBankmRNA HY010169.1 - 1, 2 223 Chr. 10 - 87238758 - 87342455 [-] on Build GRCh38
          GenBankmRNA HY124799.1 - 1, 2 115 Chr. 10 - 87216064 - 87342347 [-] on Build GRCh38
          GenBankmRNA BG748770.1 - 1, 2 327 Chr. 10 - 79766290 - 79826477 [-] on Build GRCh38
          GenBankmRNA BE742363.1 - 222 Chr. - - [-] on Build
          GenBankmRNA DB456680.1 - 1, 2 252 Chr. 10 - 87246163 - 87342485 [-] on Build GRCh38
          GenBankmRNA HY151352.1 - 1, 2 245 Chr. 10 - 87208482 - 87342477 [-] on Build GRCh38
          GenBankmRNA DA571282.1 - 1, 2 167 Chr. 10 - 87288087 - 87342400 [-] on Build GRCh38
          GenBankmRNA DA829196.1 - 1, 2 122 Chr. 10 - 87287993 - 87342355 [-] on Build GRCh38
          GenBankmRNA DA258253.1 - 1, 2 144 Chr. 10 - 87216011 - 87342377 [-] on Build GRCh38
          GenBankmRNA DA259512.1 - 1, 2 150 Chr. 10 - 87202973 - 87342383 [-] on Build GRCh38
          GenBankmRNA DA551755.1 - 1, 2 110 Chr. 10 - 87245749 - 87342343 [-] on Build GRCh38
          GenBankmRNA DB263965.1 - 1, 2 137 Chr. 10 - 87206378 - 87342370 [-] on Build GRCh38
          GenBankmRNA DA009741.1 - 1, 2 142 Chr. 10 - 87245758 - 87342375 [-] on Build GRCh38
          GenBankmRNA HY055033.1 - 1, 2 223 Chr. 10 - 87288436 - 87342455 [-] on Build GRCh38
          GenBankmRNA BM053142.1 - 1, 2 335 Chr. 10 - 79808759 - 79826515 [-] on Build GRCh38
          GenBankmRNA BF437950.1 - 1, 2 152 Chr. 10 - 79801984 - 79826341 [-] on Build GRCh38
          GenBankmRNA AW204120.1 - 1, 2 202 Chr. 10 - 79804494 - 79826391 [-] on Build GRCh38
          GenBankmRNA DA775058.1 - 1, 2 145 Chr. 10 - 87215939 - 87342377 [-] on Build GRCh38
          GenBankmRNA HY157097.1 - 1, 2 115 Chr. 10 - 87216111 - 87342347 [-] on Build GRCh38
          GenBankmRNA CN279212.1 - 1, 2 247 Chr. 10 - 79804510 - 79826427 [-] on Build GRCh38
          GenBankmRNA BG256210.1 - 194 Chr. - - [-] on Build

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-bqzg7:80/100.66.78.82:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0