Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LOC100132057 SIRNA Assays ›
          Gene Symbol:
          LOC100132057
          Gene Name:
          phosphodiesterase 4D interacting protein pseudogene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 148402483 - 148432545 on Build GRCh38
          UniGene ID:
          Hs.585189
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n270452

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LOC100132057
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_111930.1 1, 2 182
          GenBank mRNA AK311108.1 - 1, 2 165
          BX464028.2 - 1, 2 151
          DB107560.1 - 1, 2 179
          DB442034.1 - 1, 2 182
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA CR746295.1 - 1, 2 113 Chr. 1 - 148402553 - 148427882 [+] on Build GRCh38
          GenBankmRNA DA810129.1 - 1, 2 157 Chr. 1 - 148402508 - 148420764 [+] on Build GRCh38
          GenBankmRNA HY060671.1 - 1, 2 131 Chr. 1 - 148402534 - 148424710 [+] on Build GRCh38
          GenBankmRNA BI912967.1 - 1, 2 212 Chr. 1 - 148402453 - 148421128 [+] on Build GRCh38
          GenBankmRNA R34603.1 - 110 Chr. - - [-] on Build
          GenBankmRNA BM928261.1 - 1, 2 66 Chr. 1 - 148402599 - 148432302 [+] on Build GRCh38
          GenBankmRNA DA097547.1 - 1, 2 165 Chr. 1 - 148402500 - 148420762 [+] on Build GRCh38
          GenBankmRNA BX442013.2 - 1, 2 152 Chr. 1 - 148402514 - 148428828 [+] on Build GRCh38
          GenBankmRNA R31379.1 - 110 Chr. - - [-] on Build
          GenBankmRNA DA157610.1 - 1, 2 155 Chr. 1 - 148402510 - 148420985 [+] on Build GRCh38
          GenBankmRNA AV711778.1 - 1, 2 157 Chr. 1 - 148402508 - 148432124 [+] on Build GRCh38
          GenBankmRNA HY081465.1 - 1, 2 120 Chr. 1 - 148402546 - 148425482 [+] on Build GRCh38
          GenBankmRNA DA521846.1 - 1, 2 159 Chr. 1 - 148402506 - 148420994 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline