Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ZWILCH SIRNA Assays ›
          Gene Symbol:
          ZWILCH
          Gene Name:
          zwilch kinetochore protein
          Gene Aliases:
          KNTC1AP, hZwilch
          Chromosome Location:
          Chr.15: 66505083 - 66549485 on Build GRCh38
          UniGene ID:
          Hs.21331
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n272550

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ZWILCH
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001287821.1 NP_001274750.1 2 334
          NM_001287822.1 NP_001274751.1 2 334
          GenBank mRNA AK122787.1 - 2 122
          BC090041.1 - 2 83 6472990
          BX640701.1 - 2 301
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BP230336.1 - 2 91 Chr. 15 - 66505326 - 66520657 [+] on Build GRCh38
          GenBankmRNA BG773076.1 - 2 117 Chr. 15 - 66505305 - 66520643 [+] on Build GRCh38
          GenBankmRNA DB072733.1 - 2 291 Chr. 15 - 66505126 - 66515548 [+] on Build GRCh38
          GenBankmRNA BG037081.1 - 331 Chr. - - [-] on Build
          GenBankmRNA DB079543.1 - 2 122 Chr. 15 - 66505295 - 66519001 [+] on Build GRCh38
          GenBankmRNA DB457173.1 - 2 103 Chr. 15 - 66505314 - 66518930 [+] on Build GRCh38
          GenBankmRNA DB457086.1 - 2 89 Chr. 15 - 66505328 - 66518945 [+] on Build GRCh38
          GenBankmRNA DB078094.1 - 2 117 Chr. 15 - 66505300 - 66518993 [+] on Build GRCh38
          GenBankmRNA DB036780.1 - 2 117 Chr. 15 - 66505300 - 66518999 [+] on Build GRCh38
          GenBankmRNA BI561980.1 - 2 103 Chr. 15 - 66505320 - 66519077 [+] on Build GRCh38
          GenBankmRNA BG718708.1 - 2 327 Chr. 15 - 66505089 - 66518947 [+] on Build GRCh38
          GenBankmRNA DB442406.1 - 2 124 Chr. 15 - 66505295 - 66515613 [+] on Build GRCh38
          GenBankmRNA DA740997.1 - 2 78 Chr. 15 - 66505340 - 66518976 [+] on Build GRCh38
          GenBankmRNA DB100639.1 - 2 286 Chr. 15 - 66505131 - 66515625 [+] on Build GRCh38
          GenBankmRNA DB051540.1 - 2 117 Chr. 15 - 66505300 - 66518990 [+] on Build GRCh38
          GenBankmRNA BX441738.2 - 2 66 Chr. 15 - 66505350 - 66520646 [+] on Build GRCh38
          GenBankmRNA DB454015.1 - 2 122 Chr. 15 - 66505295 - 66518912 [+] on Build GRCh38
          GenBankmRNA DB028207.1 - 2 118 Chr. 15 - 66505299 - 66518954 [+] on Build GRCh38
          GenBankmRNA DB462505.1 - 2 104 Chr. 15 - 66505314 - 66518926 [+] on Build GRCh38
          GenBankmRNA DA189358.1 - 2 285 Chr. 15 - 66505135 - 66515612 [+] on Build GRCh38
          GenBankmRNA BI560087.1 - 2 108 Chr. 15 - 66505313 - 66519061 [+] on Build GRCh38
          GenBankmRNA DB094185.1 - 2 122 Chr. 15 - 66505295 - 66518962 [+] on Build GRCh38
          GenBankmRNA BQ938079.1 - 2 81 Chr. 15 - 66505336 - 66523750 [+] on Build GRCh38
          GenBankmRNA DB444216.1 - 2 89 Chr. 15 - 66505329 - 66515650 [+] on Build GRCh38
          GenBankmRNA DB442087.1 - 2 90 Chr. 15 - 66505328 - 66515650 [+] on Build GRCh38
          GenBankmRNA HY029861.1 - 2 118 Chr. 15 - 66505300 - 66515631 [+] on Build GRCh38
          GenBankmRNA DB085281.1 - 2 91 Chr. 15 - 66505326 - 66519010 [+] on Build GRCh38
          GenBankmRNA DB075492.1 - 2 122 Chr. 15 - 66505295 - 66519003 [+] on Build GRCh38
          GenBankmRNA DB452796.1 - 1 39 Chr. 15 - 66508744 - 66518976 [+] on Build GRCh38
          GenBankmRNA DB445221.1 - 2 119 Chr. 15 - 66505300 - 66515622 [+] on Build GRCh38
          GenBankmRNA DB280397.1 - 2 257 Chr. 15 - 66505161 - 66514349 [+] on Build GRCh38
          GenBankmRNA DB455839.1 - 2 89 Chr. 15 - 66505328 - 66518930 [+] on Build GRCh38
          GenBankmRNA BI831378.1 - 2 89 Chr. 15 - 66505328 - 66521155 [+] on Build GRCh38
          GenBankmRNA DB074483.1 - 2 93 Chr. 15 - 66505325 - 66518992 [+] on Build GRCh38
          GenBankmRNA DB026091.1 - 2 122 Chr. 15 - 66505295 - 66518974 [+] on Build GRCh38
          GenBankmRNA DB080227.1 - 2 89 Chr. 15 - 66505329 - 66519019 [+] on Build GRCh38
          GenBankmRNA BG720559.1 - 2 114 Chr. 15 - 66505306 - 66519079 [+] on Build GRCh38
          GenBankmRNA DB458643.1 - 2 89 Chr. 15 - 66505328 - 66518929 [+] on Build GRCh38
          GenBankmRNA DA003230.1 - 2 115 Chr. 15 - 66505302 - 66518990 [+] on Build GRCh38
          GenBankmRNA BQ223339.1 - 72 Chr. - - [-] on Build
          GenBankmRNA BG719511.1 - 2 121 Chr. 15 - 66505303 - 66520647 [+] on Build GRCh38
          GenBankmRNA DB121865.1 - 2 89 Chr. 15 - 66505328 - 66518999 [+] on Build GRCh38
          GenBankmRNA DB459706.1 - 1 39 Chr. 15 - 66508744 - 66518952 [+] on Build GRCh38
          GenBankmRNA DB072717.1 - 2 152 Chr. 15 - 66505265 - 66518926 [+] on Build GRCh38
          GenBankmRNA DB038415.1 - 2 287 Chr. 15 - 66505131 - 66515597 [+] on Build GRCh38
          GenBankmRNA BG772880.1 - 2 117 Chr. 15 - 66505305 - 66521109 [+] on Build GRCh38
          GenBankmRNA HY011977.1 - 2 118 Chr. 15 - 66505301 - 66518959 [+] on Build GRCh38
          GenBankmRNA DB078682.1 - 2 122 Chr. 15 - 66505295 - 66518997 [+] on Build GRCh38
          GenBankmRNA BG718028.1 - 2 122 Chr. 15 - 66505300 - 66518974 [+] on Build GRCh38
          GenBankmRNA DB458798.1 - 2 117 Chr. 15 - 66505300 - 66515644 [+] on Build GRCh38
          GenBankmRNA HY005961.1 - 2 110 Chr. 15 - 66505308 - 66518925 [+] on Build GRCh38
          GenBankmRNA DB451512.1 - 2 122 Chr. 15 - 66505295 - 66518915 [+] on Build GRCh38
          GenBankmRNA BM172304.1 - 2 117 Chr. 15 - 66505305 - 66519053 [+] on Build GRCh38
          GenBankmRNA DA166613.1 - 2 91 Chr. 15 - 66505326 - 66518957 [+] on Build GRCh38
          GenBankmRNA BG389949.1 - 50 Chr. - - [-] on Build
          GenBankmRNA DB075562.1 - 2 122 Chr. 15 - 66505295 - 66518997 [+] on Build GRCh38
          GenBankmRNA BG037068.1 - 121 Chr. - - [-] on Build
          GenBankmRNA DB082591.1 - 2 122 Chr. 15 - 66505295 - 66518967 [+] on Build GRCh38
          GenBankmRNA DB099221.1 - 2 108 Chr. 15 - 66505309 - 66519033 [+] on Build GRCh38
          GenBankmRNA DB058761.1 - 2 117 Chr. 15 - 66505300 - 66518980 [+] on Build GRCh38
          GenBankmRNA DB076740.1 - 2 117 Chr. 15 - 66505300 - 66518957 [+] on Build GRCh38
          GenBankmRNA DB141816.1 - 2 89 Chr. 15 - 66505328 - 66518983 [+] on Build GRCh38
          GenBankmRNA BG772087.1 - 2 329 Chr. 15 - 66505092 - 66518956 [+] on Build GRCh38
          GenBankmRNA HY010286.1 - 2 123 Chr. 15 - 66505295 - 66518922 [+] on Build GRCh38
          GenBankmRNA DB072565.1 - 2 300 Chr. 15 - 66505117 - 66514083 [+] on Build GRCh38
          GenBankmRNA DB099232.1 - 2 91 Chr. 15 - 66505326 - 66519044 [+] on Build GRCh38
          GenBankmRNA HY036298.1 - 2 123 Chr. 15 - 66505295 - 66515640 [+] on Build GRCh38
          GenBankmRNA DB155700.1 - 2 117 Chr. 15 - 66505300 - 66519018 [+] on Build GRCh38
          GenBankmRNA BG772339.1 - 2 111 Chr. 15 - 66505312 - 66519079 [+] on Build GRCh38
          GenBankmRNA DB080223.1 - 2 114 Chr. 15 - 66505303 - 66518995 [+] on Build GRCh38
          GenBankmRNA DB072779.1 - 2 286 Chr. 15 - 66505131 - 66515569 [+] on Build GRCh38
          GenBankmRNA DA501613.1 - 2 153 Chr. 15 - 66505265 - 66518939 [+] on Build GRCh38
          GenBankmRNA DB089252.1 - 2 122 Chr. 15 - 66505295 - 66518967 [+] on Build GRCh38
          GenBankmRNA HY183276.1 - 2 89 Chr. 15 - 66505328 - 66518956 [+] on Build GRCh38
          GenBankmRNA BI562310.1 - 2 111 Chr. 15 - 66505309 - 66519063 [+] on Build GRCh38
          GenBankmRNA DB100359.1 - 2 140 Chr. 15 - 66505277 - 66518989 [+] on Build GRCh38
          GenBankmRNA BI560051.1 - 2 121 Chr. 15 - 66505300 - 66520641 [+] on Build GRCh38
          GenBankmRNA DA410992.1 - 2 90 Chr. 15 - 66505327 - 66519022 [+] on Build GRCh38
          GenBankmRNA DA690141.1 - 2 280 Chr. 15 - 66505140 - 66515579 [+] on Build GRCh38
          GenBankmRNA BG721310.1 - 2 122 Chr. 15 - 66505300 - 66519079 [+] on Build GRCh38
          GenBankmRNA BI561060.1 - 2 126 Chr. 15 - 66505296 - 66520656 [+] on Build GRCh38
          GenBankmRNA DB028815.1 - 2 324 Chr. 15 - 66505093 - 66515581 [+] on Build GRCh38
          GenBankmRNA DB025287.1 - 2 108 Chr. 15 - 66505309 - 66518985 [+] on Build GRCh38
          GenBankmRNA BI461145.1 - 2 91 Chr. 15 - 66505331 - 66519084 [+] on Build GRCh38
          GenBankmRNA DB020138.1 - 2 117 Chr. 15 - 66505300 - 66518977 [+] on Build GRCh38
          GenBankmRNA HY055507.1 - 2 118 Chr. 15 - 66505300 - 66515642 [+] on Build GRCh38
          GenBankmRNA DB264903.1 - 2 91 Chr. 15 - 66505326 - 66519040 [+] on Build GRCh38

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          sister chromatid cohesion
          mitotic nuclear division
          mitotic cell cycle checkpoint
          cell division
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-64twl:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0