Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ZWILCH SIRNA Assays ›
          Gene Symbol:
          ZWILCH
          Gene Name:
          zwilch kinetochore protein
          Gene Aliases:
          KNTC1AP, hZwilch
          Chromosome Location:
          Chr.15: 66505083 - 66549485 on Build GRCh38
          UniGene ID:
          Hs.21331
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n272551

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ZWILCH
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001287821.1 NP_001274750.1 2 398
          NM_001287822.1 NP_001274751.1 2 398
          GenBank mRNA AK122787.1 - 2 186
          BC090041.1 - 2 147 6472990
          BX640701.1 - 2 365
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DB072733.1 - 2 355 Chr. 15 - 66505126 - 66515548 [+] on Build GRCh38
          GenBankmRNA DB079543.1 - 2 186 Chr. 15 - 66505295 - 66519001 [+] on Build GRCh38
          GenBankmRNA DB457086.1 - 2 153 Chr. 15 - 66505328 - 66518945 [+] on Build GRCh38
          GenBankmRNA DB078094.1 - 2 182 Chr. 15 - 66505300 - 66518993 [+] on Build GRCh38
          GenBankmRNA BI561980.1 - 2 167 Chr. 15 - 66505320 - 66519077 [+] on Build GRCh38
          GenBankmRNA DA740997.1 - 2 142 Chr. 15 - 66505340 - 66518976 [+] on Build GRCh38
          GenBankmRNA DB100639.1 - 2 350 Chr. 15 - 66505131 - 66515625 [+] on Build GRCh38
          GenBankmRNA BX441738.2 - 2 130 Chr. 15 - 66505350 - 66520646 [+] on Build GRCh38
          GenBankmRNA DB094185.1 - 2 187 Chr. 15 - 66505295 - 66518962 [+] on Build GRCh38
          GenBankmRNA DB444216.1 - 2 153 Chr. 15 - 66505329 - 66515650 [+] on Build GRCh38
          GenBankmRNA BQ938079.1 - 2 145 Chr. 15 - 66505336 - 66523750 [+] on Build GRCh38
          GenBankmRNA DB085281.1 - 2 155 Chr. 15 - 66505326 - 66519010 [+] on Build GRCh38
          GenBankmRNA DB455839.1 - 2 153 Chr. 15 - 66505328 - 66518930 [+] on Build GRCh38
          GenBankmRNA DB074483.1 - 2 157 Chr. 15 - 66505325 - 66518992 [+] on Build GRCh38
          GenBankmRNA DB026091.1 - 2 187 Chr. 15 - 66505295 - 66518974 [+] on Build GRCh38
          GenBankmRNA DB080227.1 - 2 153 Chr. 15 - 66505329 - 66519019 [+] on Build GRCh38
          GenBankmRNA BG720559.1 - 2 178 Chr. 15 - 66505306 - 66519079 [+] on Build GRCh38
          GenBankmRNA DB458643.1 - 2 153 Chr. 15 - 66505328 - 66518929 [+] on Build GRCh38
          GenBankmRNA BQ223339.1 - 136 Chr. - - [-] on Build
          GenBankmRNA DB459706.1 - 1 103 Chr. 15 - 66508744 - 66518952 [+] on Build GRCh38
          GenBankmRNA DB121865.1 - 2 153 Chr. 15 - 66505328 - 66518999 [+] on Build GRCh38
          GenBankmRNA BG719511.1 - 2 185 Chr. 15 - 66505303 - 66520647 [+] on Build GRCh38
          GenBankmRNA DB038415.1 - 2 351 Chr. 15 - 66505131 - 66515597 [+] on Build GRCh38
          GenBankmRNA HY011977.1 - 2 182 Chr. 15 - 66505301 - 66518959 [+] on Build GRCh38
          GenBankmRNA HY005961.1 - 2 174 Chr. 15 - 66505308 - 66518925 [+] on Build GRCh38
          GenBankmRNA DB451512.1 - 2 186 Chr. 15 - 66505295 - 66518915 [+] on Build GRCh38
          GenBankmRNA DA166613.1 - 2 155 Chr. 15 - 66505326 - 66518957 [+] on Build GRCh38
          GenBankmRNA DB075562.1 - 2 187 Chr. 15 - 66505295 - 66518997 [+] on Build GRCh38
          GenBankmRNA DB099221.1 - 2 172 Chr. 15 - 66505309 - 66519033 [+] on Build GRCh38
          GenBankmRNA BG772087.1 - 2 393 Chr. 15 - 66505092 - 66518956 [+] on Build GRCh38
          GenBankmRNA HY010286.1 - 2 187 Chr. 15 - 66505295 - 66518922 [+] on Build GRCh38
          GenBankmRNA DB072565.1 - 2 364 Chr. 15 - 66505117 - 66514083 [+] on Build GRCh38
          GenBankmRNA DB099232.1 - 2 155 Chr. 15 - 66505326 - 66519044 [+] on Build GRCh38
          GenBankmRNA HY036298.1 - 2 187 Chr. 15 - 66505295 - 66515640 [+] on Build GRCh38
          GenBankmRNA DA501613.1 - 2 217 Chr. 15 - 66505265 - 66518939 [+] on Build GRCh38
          GenBankmRNA BE246190.1 - 2 155 Chr. 15 - 66505327 - 66515637 [+] on Build GRCh38
          GenBankmRNA DB100359.1 - 2 205 Chr. 15 - 66505277 - 66518989 [+] on Build GRCh38
          GenBankmRNA DB086248.1 - 2 216 Chr. 15 - 66505267 - 66518956 [+] on Build GRCh38
          GenBankmRNA DB093569.1 - 2 398 Chr. 15 - 66505083 - 66514083 [+] on Build GRCh38
          GenBankmRNA DA410992.1 - 2 154 Chr. 15 - 66505327 - 66519022 [+] on Build GRCh38
          GenBankmRNA BG721310.1 - 2 186 Chr. 15 - 66505300 - 66519079 [+] on Build GRCh38
          GenBankmRNA DB025287.1 - 2 172 Chr. 15 - 66505309 - 66518985 [+] on Build GRCh38
          GenBankmRNA DB073950.1 - 2 183 Chr. 15 - 66505300 - 66518960 [+] on Build GRCh38
          GenBankmRNA HY055507.1 - 2 182 Chr. 15 - 66505300 - 66515642 [+] on Build GRCh38
          GenBankmRNA DB264903.1 - 2 155 Chr. 15 - 66505326 - 66519040 [+] on Build GRCh38
          GenBankmRNA BG773076.1 - 2 181 Chr. 15 - 66505305 - 66520643 [+] on Build GRCh38
          GenBankmRNA BG037081.1 - 395 Chr. - - [-] on Build
          GenBankmRNA BP230336.1 - 2 155 Chr. 15 - 66505326 - 66520657 [+] on Build GRCh38
          GenBankmRNA DB457173.1 - 2 167 Chr. 15 - 66505314 - 66518930 [+] on Build GRCh38
          GenBankmRNA DB036780.1 - 2 181 Chr. 15 - 66505300 - 66518999 [+] on Build GRCh38
          GenBankmRNA BG718708.1 - 2 391 Chr. 15 - 66505089 - 66518947 [+] on Build GRCh38
          GenBankmRNA DB442406.1 - 2 188 Chr. 15 - 66505295 - 66515613 [+] on Build GRCh38
          GenBankmRNA BE244105.1 - 2 155 Chr. 15 - 66505327 - 66515562 [+] on Build GRCh38
          GenBankmRNA DB051540.1 - 2 182 Chr. 15 - 66505300 - 66518990 [+] on Build GRCh38
          GenBankmRNA DB454015.1 - 2 186 Chr. 15 - 66505295 - 66518912 [+] on Build GRCh38
          GenBankmRNA DB028207.1 - 2 182 Chr. 15 - 66505299 - 66518954 [+] on Build GRCh38
          GenBankmRNA DB462505.1 - 2 168 Chr. 15 - 66505314 - 66518926 [+] on Build GRCh38
          GenBankmRNA DA189358.1 - 2 349 Chr. 15 - 66505135 - 66515612 [+] on Build GRCh38
          GenBankmRNA BI560087.1 - 2 172 Chr. 15 - 66505313 - 66519061 [+] on Build GRCh38
          GenBankmRNA HY029861.1 - 2 181 Chr. 15 - 66505300 - 66515631 [+] on Build GRCh38
          GenBankmRNA DB442087.1 - 2 154 Chr. 15 - 66505328 - 66515650 [+] on Build GRCh38
          GenBankmRNA DB075492.1 - 2 187 Chr. 15 - 66505295 - 66519003 [+] on Build GRCh38
          GenBankmRNA DB445221.1 - 2 183 Chr. 15 - 66505300 - 66515622 [+] on Build GRCh38
          GenBankmRNA DB452796.1 - 1 103 Chr. 15 - 66508744 - 66518976 [+] on Build GRCh38
          GenBankmRNA DB280397.1 - 2 321 Chr. 15 - 66505161 - 66514349 [+] on Build GRCh38
          GenBankmRNA BI831378.1 - 2 153 Chr. 15 - 66505328 - 66521155 [+] on Build GRCh38
          GenBankmRNA DA003230.1 - 2 180 Chr. 15 - 66505302 - 66518990 [+] on Build GRCh38
          GenBankmRNA DB072717.1 - 2 216 Chr. 15 - 66505265 - 66518926 [+] on Build GRCh38
          GenBankmRNA BG772880.1 - 2 181 Chr. 15 - 66505305 - 66521109 [+] on Build GRCh38
          GenBankmRNA DB078682.1 - 2 187 Chr. 15 - 66505295 - 66518997 [+] on Build GRCh38
          GenBankmRNA BG718028.1 - 2 187 Chr. 15 - 66505300 - 66518974 [+] on Build GRCh38
          GenBankmRNA DB458798.1 - 2 181 Chr. 15 - 66505300 - 66515644 [+] on Build GRCh38
          GenBankmRNA BM172304.1 - 2 181 Chr. 15 - 66505305 - 66519053 [+] on Build GRCh38
          GenBankmRNA BG389949.1 - 114 Chr. - - [-] on Build
          GenBankmRNA BG037068.1 - 185 Chr. - - [-] on Build
          GenBankmRNA DB082591.1 - 2 187 Chr. 15 - 66505295 - 66518967 [+] on Build GRCh38
          GenBankmRNA DB093601.1 - 2 398 Chr. 15 - 66505083 - 66514083 [+] on Build GRCh38
          GenBankmRNA DB058761.1 - 2 182 Chr. 15 - 66505300 - 66518980 [+] on Build GRCh38
          GenBankmRNA DB076740.1 - 2 182 Chr. 15 - 66505300 - 66518957 [+] on Build GRCh38
          GenBankmRNA DB141816.1 - 2 153 Chr. 15 - 66505328 - 66518983 [+] on Build GRCh38
          GenBankmRNA DB155700.1 - 2 182 Chr. 15 - 66505300 - 66519018 [+] on Build GRCh38
          GenBankmRNA DA805639.1 - 2 182 Chr. 15 - 66505300 - 66514522 [+] on Build GRCh38
          GenBankmRNA BG772339.1 - 2 175 Chr. 15 - 66505312 - 66519079 [+] on Build GRCh38
          GenBankmRNA DB080223.1 - 2 178 Chr. 15 - 66505303 - 66518995 [+] on Build GRCh38
          GenBankmRNA DB072779.1 - 2 350 Chr. 15 - 66505131 - 66515569 [+] on Build GRCh38
          GenBankmRNA HY183276.1 - 2 153 Chr. 15 - 66505328 - 66518956 [+] on Build GRCh38
          GenBankmRNA DB089252.1 - 2 187 Chr. 15 - 66505295 - 66518967 [+] on Build GRCh38
          GenBankmRNA BI562310.1 - 2 175 Chr. 15 - 66505309 - 66519063 [+] on Build GRCh38
          GenBankmRNA BI560051.1 - 2 185 Chr. 15 - 66505300 - 66520641 [+] on Build GRCh38
          GenBankmRNA DA690141.1 - 2 344 Chr. 15 - 66505140 - 66515579 [+] on Build GRCh38
          GenBankmRNA BI561060.1 - 2 190 Chr. 15 - 66505296 - 66520656 [+] on Build GRCh38
          GenBankmRNA DB028815.1 - 2 388 Chr. 15 - 66505093 - 66515581 [+] on Build GRCh38
          GenBankmRNA BI461145.1 - 2 155 Chr. 15 - 66505331 - 66519084 [+] on Build GRCh38
          GenBankmRNA DB020138.1 - 2 182 Chr. 15 - 66505300 - 66518977 [+] on Build GRCh38
          GenBankmRNA DB478511.1 - 2 181 Chr. 15 - 66505300 - 66515647 [+] on Build GRCh38

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          sister chromatid cohesion
          mitotic nuclear division
          mitotic cell cycle checkpoint
          cell division
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0