Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MEG3 SIRNA Assays ›
          Gene Symbol:
          MEG3
          Gene Name:
          maternally expressed 3 (non-protein coding)
          Gene Aliases:
          FP504, GTL2, LINC00023, NCRNA00023, PRO0518, PRO2160, onco-lncRNA-83, prebp1
          Chromosome Location:
          Chr.14: 100826108 - 100861026 on Build GRCh38
          UniGene ID:
          Hs.654863
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n272559

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MEG3
          Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002766.2
          3 883
          NR_003530.2
          3 883
          NR_003531.3
          3 859
          NR_033358.1
          3 883
          NR_033359.1
          3 883
          NR_033360.1
          3 883
          NR_046464.1
          3 883
          NR_046465.2
          3 883
          NR_046466.1
          3 883
          NR_046467.1
          3 883
          NR_046469.1
          3 883
          NR_046470.2
          3 883
          NR_046471.1
          3 883
          NR_046472.1
          3 883
          NR_046473.1
          3 883
          GenBank mRNA AB032607.1 -
          3 870
          AF052114.1 -
          1 625
          AY314975.1 - LIT3321
          MEG3
          3 859
          BC023543.2 -
          3 858
          BC092509.1 -
          1 78
          GQ183494.1 -
          3 875
          GQ183495.1 -
          3 875
          GQ183496.1 -
          3 875
          GQ183497.1 -
          3 875
          GQ183498.1 -
          3 875
          GQ183499.1 -
          3 875
          GQ183500.1 -
          3 875
          GQ183501.1 -
          3 875
          GQ183502.1 -
          3 875
          GQ183503.1 -
          3 875
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BF932705.1 - 127 Chr. - - [-] on Build
          GenBankmRNA H58895.1 - 88 Chr. - - [-] on Build
          GenBankmRNA BX326323.2 - 1 68 Chr. 14 - 100829630 - 100861050 [+] on Build GRCh38
          GenBankmRNA BY796127.2 - 1 252 Chr. 14 - 100829446 - 100845508 [+] on Build GRCh38
          GenBankmRNA BG703541.1 - 1 83 Chr. 14 - 100829615 - 100846219 [+] on Build GRCh38
          GenBankmRNA CA943189.1 - 1 177 Chr. 14 - 100829522 - 100845520 [+] on Build GRCh38
          GenBankmRNA BX346951.1 - 1 189 Chr. 14 - 100829508 - 100860951 [+] on Build GRCh38
          GenBankmRNA BX429954.2 - 1 384 Chr. 14 - 100829354 - 100845518 [+] on Build GRCh38
          GenBankmRNA BX408784.2 - 1 122 Chr. 14 - 100829579 - 100861025 [+] on Build GRCh38
          GenBankmRNA AI174720.1 - 1 301 Chr. 14 - 100829397 - 100836304 [+] on Build GRCh38
          GenBankmRNA BX368158.1 - 1 325 Chr. 14 - 100829371 - 100860859 [+] on Build GRCh38
          GenBankmRNA H59650.1 - 78 Chr. - - [-] on Build
          GenBankmRNA BX408697.2 - 1 57 Chr. 14 - 100829658 - 100861025 [+] on Build GRCh38
          GenBankmRNA BX367746.1 - 1 290 Chr. 14 - 100829409 - 100860888 [+] on Build GRCh38
          GenBankmRNA BX450276.1 - 1 214 Chr. 14 - 100829484 - 100846142 [+] on Build GRCh38
          GenBankmRNA BX408219.1 - 1 190 Chr. 14 - 100829507 - 100860967 [+] on Build GRCh38
          GenBankmRNA DA361098.1 - 1 62 Chr. 14 - 100829636 - 100845997 [+] on Build GRCh38
          GenBankmRNA DA083332.1 - 1 185 Chr. 14 - 100829513 - 100836289 [+] on Build GRCh38
          GenBankmRNA CB154926.1 - 1 130 Chr. 14 - 100829568 - 100860741 [+] on Build GRCh38
          GenBankmRNA AV723467.1 - 1 39 Chr. 14 - 100829658 - 100860731 [+] on Build GRCh38
          GenBankmRNA BU075431.1 - 1 41 Chr. 14 - 100829679 - 100860862 [+] on Build GRCh38
          GenBankmRNA BX388604.2 - 1 235 Chr. 14 - 100829461 - 100836290 [+] on Build GRCh38
          GenBankmRNA BX429896.2 - 1 73 Chr. 14 - 100829639 - 100846333 [+] on Build GRCh38
          GenBankmRNA AV753036.1 - 224 Chr. - - [-] on Build
          GenBankmRNA BX422168.2 - 1 339 Chr. 14 - 100829359 - 100860947 [+] on Build GRCh38
          GenBankmRNA BX347225.2 - 1 141 Chr. 14 - 100829557 - 100836299 [+] on Build GRCh38
          GenBankmRNA BX408226.1 - 1 207 Chr. 14 - 100829491 - 100860979 [+] on Build GRCh38
          GenBankmRNA H71305.1 - 1 121 Chr. 14 - 100829576 - 100832632 [+] on Build GRCh38
          GenBankmRNA BX429869.2 - 1 71 Chr. 14 - 100829629 - 100861023 [+] on Build GRCh38
          GenBankmRNA BX408189.1 - 1 236 Chr. 14 - 100829460 - 100860925 [+] on Build GRCh38
          GenBankmRNA AU309923.1 - 1 59 Chr. 14 - 100829639 - 100846016 [+] on Build GRCh38
          GenBankmRNA BU580862.1 - 1 140 Chr. 14 - 100829558 - 100860756 [+] on Build GRCh38
          GenBankmRNA BX450687.2 - 1 114 Chr. 14 - 100829584 - 100860988 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-4pjxj:80/100.66.79.163:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0