Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MEG3 SIRNA Assays ›
          Gene Symbol:
          MEG3
          Gene Name:
          maternally expressed 3 (non-protein coding)
          Gene Aliases:
          FP504, GTL2, LINC00023, NCRNA00023, PRO0518, PRO2160, onco-lncRNA-83, prebp1
          Chromosome Location:
          Chr.14: 100826108 - 100861026 on Build GRCh38
          UniGene ID:
          Hs.654863
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n272560

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MEG3
          Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002766.2
          3 731
          NR_003530.2
          3 731
          NR_003531.3
          3 707
          NR_033358.1
          3 731
          NR_033359.1
          3 731
          NR_033360.1
          3 731
          NR_046464.1
          3 731
          NR_046465.2
          3 731
          NR_046466.1
          3 731
          NR_046467.1
          3 731
          NR_046469.1
          3 731
          NR_046470.2
          3 731
          NR_046471.1
          3 731
          NR_046472.1
          3 731
          NR_046473.1
          3 731
          GenBank mRNA AB032607.1 -
          3 718
          AF052114.1 -
          1 473
          AY314975.1 - LIT3321
          MEG3
          3 707
          BC023543.2 -
          3 706
          GQ183494.1 -
          3 723
          GQ183495.1 -
          3 723
          GQ183496.1 -
          3 723
          GQ183497.1 -
          3 723
          GQ183498.1 -
          3 723
          GQ183499.1 -
          3 723
          GQ183500.1 -
          3 723
          GQ183501.1 -
          3 723
          GQ183502.1 -
          3 723
          GQ183503.1 -
          3 723
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA083332.1 - 1 33 Chr. 14 - 100829513 - 100836289 [+] on Build GRCh38
          GenBankmRNA AV727351.1 - 1 221 Chr. 14 - 100829325 - 100845483 [+] on Build GRCh38
          GenBankmRNA BX388604.2 - 1 83 Chr. 14 - 100829461 - 100836290 [+] on Build GRCh38
          GenBankmRNA AV753036.1 - 72 Chr. - - [-] on Build
          GenBankmRNA BY796127.2 - 1 100 Chr. 14 - 100829446 - 100845508 [+] on Build GRCh38
          GenBankmRNA CA943189.1 - 1 25 Chr. 14 - 100829522 - 100845520 [+] on Build GRCh38
          GenBankmRNA BX422168.2 - 1 187 Chr. 14 - 100829359 - 100860947 [+] on Build GRCh38
          GenBankmRNA BX429954.2 - 1 232 Chr. 14 - 100829354 - 100845518 [+] on Build GRCh38
          GenBankmRNA BX346951.1 - 1 38 Chr. 14 - 100829508 - 100860951 [+] on Build GRCh38
          GenBankmRNA AI174720.1 - 1 149 Chr. 14 - 100829397 - 100836304 [+] on Build GRCh38
          GenBankmRNA BX408226.1 - 1 55 Chr. 14 - 100829491 - 100860979 [+] on Build GRCh38
          GenBankmRNA BX368158.1 - 1 173 Chr. 14 - 100829371 - 100860859 [+] on Build GRCh38
          GenBankmRNA BX367746.1 - 1 138 Chr. 14 - 100829409 - 100860888 [+] on Build GRCh38
          GenBankmRNA BX408189.1 - 1 84 Chr. 14 - 100829460 - 100860925 [+] on Build GRCh38
          GenBankmRNA BX408219.1 - 1 39 Chr. 14 - 100829507 - 100860967 [+] on Build GRCh38
          GenBankmRNA BX450276.1 - 1 62 Chr. 14 - 100829484 - 100846142 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0