Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LIMS1 SIRNA Assays ›
          Gene Symbol:
          LIMS1
          Gene Name:
          LIM zinc finger domain containing 1
          Gene Aliases:
          PINCH, PINCH-1, PINCH1
          Chromosome Location:
          Chr.2: 108534355 - 108687246 on Build GRCh38
          UniGene ID:
          Hs.597715
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n275722

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LIMS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001193482.1 NP_001180411.1 10 2332
          NM_001193483.2 NP_001180412.1 10 2477
          NM_001193484.1 NP_001180413.1 10 2453
          NM_001193485.2 NP_001180414.1 10 2682
          NM_001193488.1 NP_001180417.1 10 2400
          NM_004987.5 NP_004978.2 10 2362
          XM_005263946.1 XP_005264003.1 11 2674
          XM_005263948.3 XP_005264005.1 11 2727
          XM_005263949.1 XP_005264006.1 11 2669
          XM_005263950.1 XP_005264007.1 11 2510
          XM_005263951.4 XP_005264008.1 11 2946
          XM_017004091.1 XP_016859580.1 11 2696
          XM_017004092.1 XP_016859581.1 11 2691
          XM_017004093.1 XP_016859582.1 11 2740
          XM_017004094.1 XP_016859583.1 11 2975
          GenBank mRNA AK025661.1 - 1 664
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA132114.1 - 1 261 Chr. 2 - 108684994 - 108685557 [+] on Build GRCh38
          GenBankmRNA CD558324.1 - 184 Chr. - - [-] on Build
          GenBankmRNA BG611455.1 - 1 105 Chr. 2 - 108685150 - 108685883 [+] on Build GRCh38
          GenBankmRNA DA832058.1 - 1 570 Chr. 2 - 108684686 - 108685509 [+] on Build GRCh38
          GenBankmRNA DA829016.1 - 1 514 Chr. 2 - 108684741 - 108685536 [+] on Build GRCh38
          GenBankmRNA BU182475.1 - 1 359 Chr. 2 - 108684898 - 108685611 [+] on Build GRCh38
          GenBankmRNA DA572586.1 - 1 395 Chr. 2 - 108684860 - 108685648 [+] on Build GRCh38
          GenBankmRNA AI557249.1 - 1 99 Chr. 2 - 108685187 - 108685926 [+] on Build GRCh38
          GenBankmRNA BJ989975.1 - 1 424 Chr. 2 - 108684831 - 108685415 [+] on Build GRCh38
          GenBankmRNA CF144924.1 - 1 26 Chr. 2 - 108685230 - 108685772 [+] on Build GRCh38
          GenBankmRNA CN485296.1 - 1 424 Chr. 2 - 108684831 - 108685392 [+] on Build GRCh38
          GenBankmRNA BU429100.1 - 1 244 Chr. 2 - 108685014 - 108685516 [+] on Build GRCh38
          GenBankmRNA BU160720.1 - 1 112 Chr. 2 - 108685143 - 108685899 [+] on Build GRCh38
          GenBankmRNA CN388805.1 - 1 265 Chr. 2 - 108684995 - 108685473 [+] on Build GRCh38
          GenBankmRNA CN388802.1 - 1 423 Chr. 2 - 108684838 - 108685486 [+] on Build GRCh38
          GenBankmRNA AA496205.1 - 1 48 Chr. 2 - 108685207 - 108685746 [+] on Build GRCh38
          GenBankmRNA BU172480.1 - 1 272 Chr. 2 - 108684983 - 108685842 [+] on Build GRCh38
          GenBankmRNA CN388803.1 - 1 104 Chr. 2 - 108685151 - 108685636 [+] on Build GRCh38
          GenBankmRNA DA829418.1 - 1 347 Chr. 2 - 108684908 - 108685554 [+] on Build GRCh38
          GenBankmRNA BQ424598.1 - 1 135 Chr. 2 - 108685120 - 108685965 [+] on Build GRCh38
          GenBankmRNA CF146130.1 - 1 26 Chr. 2 - 108685230 - 108685582 [+] on Build GRCh38
          GenBankmRNA DA030619.1 - 1 131 Chr. 2 - 108685124 - 108685709 [+] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cell junction protein

          Gene Ontology Categories:

          Function(s) Process(es)

          epithelial to mesenchymal transition
          cell aging
          positive regulation of gene expression
          positive regulation of cell-substrate adhesion
          cell junction assembly
          positive regulation of GTPase activity
          establishment of protein localization
          negative regulation of transcription, DNA-templated
          regulation of epithelial cell proliferation
          positive regulation of focal adhesion assembly
          cellular response to transforming growth factor beta stimulus
          positive regulation of substrate adhesion-dependent cell spreading
          protein binding
          zinc ion binding
          protein kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0