Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LIMS1 SIRNA Assays ›
          Gene Symbol:
          LIMS1
          Gene Name:
          LIM zinc finger domain containing 1
          Gene Aliases:
          PINCH, PINCH-1, PINCH1
          Chromosome Location:
          Chr.2: 108534355 - 108687246 on Build GRCh38
          UniGene ID:
          Hs.597715
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n275725

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LIMS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001193482.1 NP_001180411.1 10 2713
          NM_001193483.2 NP_001180412.1 10 2858
          NM_001193484.1 NP_001180413.1 10 2834
          NM_001193485.2 NP_001180414.1 10 3063
          NM_001193488.1 NP_001180417.1 10 2781
          NM_004987.5 NP_004978.2 10 2743
          XM_005263946.1 XP_005264003.1 11 3055
          XM_005263948.3 XP_005264005.1 11 3108
          XM_005263949.1 XP_005264006.1 11 3050
          XM_005263950.1 XP_005264007.1 11 2891
          XM_005263951.4 XP_005264008.1 11 3327
          XM_017004091.1 XP_016859580.1 11 3077
          XM_017004092.1 XP_016859581.1 11 3072
          XM_017004093.1 XP_016859582.1 11 3121
          XM_017004094.1 XP_016859583.1 11 3356
          GenBank mRNA AK025661.1 - 1 1045
          AL110164.1 - 1 78
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA665633.1 - 1 345 Chr. 2 - 108685292 - 108685840 [+] on Build GRCh38
          GenBankmRNA CD558324.1 - 565 Chr. - - [-] on Build
          GenBankmRNA BG611455.1 - 1 484 Chr. 2 - 108685150 - 108685883 [+] on Build GRCh38
          GenBankmRNA BM786945.1 - 1 403 Chr. 2 - 108685249 - 108685866 [+] on Build GRCh38
          GenBankmRNA CF144924.1 - 1 408 Chr. 2 - 108685230 - 108685772 [+] on Build GRCh38
          GenBankmRNA BU160720.1 - 1 494 Chr. 2 - 108685143 - 108685899 [+] on Build GRCh38
          GenBankmRNA AA496205.1 - 1 429 Chr. 2 - 108685207 - 108685746 [+] on Build GRCh38
          GenBankmRNA BU172480.1 - 1 653 Chr. 2 - 108684983 - 108685842 [+] on Build GRCh38
          GenBankmRNA BQ424598.1 - 1 516 Chr. 2 - 108685120 - 108685965 [+] on Build GRCh38
          GenBankmRNA DA030619.1 - 1 512 Chr. 2 - 108685124 - 108685709 [+] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cell junction protein

          Gene Ontology Categories:

          Function(s) Process(es)

          epithelial to mesenchymal transition
          cell aging
          positive regulation of gene expression
          positive regulation of cell-substrate adhesion
          cell junction assembly
          positive regulation of GTPase activity
          establishment of protein localization
          negative regulation of transcription, DNA-templated
          regulation of epithelial cell proliferation
          positive regulation of focal adhesion assembly
          cellular response to transforming growth factor beta stimulus
          positive regulation of substrate adhesion-dependent cell spreading
          protein binding
          zinc ion binding
          protein kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0