Search
Search

CDS2| Gene Symbol: CDS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003818.3 | NP_003809.1 | 13 | 3745 | ||
| XM_006723660.1 | XP_006723723.1 | 13 | 3756 | |||
| XM_011529390.1 | XP_011527692.1 | 14 | 4068 | |||
| GenBank mRNA | AK130193.1 | - | 1 | 629 | ||