Search
Search

MAPK10| Gene Symbol: MAPK10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318067.1 | NP_001304996.1 | 13 | 5580 | ||
| NM_001318068.1 | NP_001304997.1 | 13 | 5675 | |||
| NM_001318069.1 | NP_001304998.1 | 14 | 6145 | |||
| NM_002753.4 | NP_002744.1 | 14 | 5686 | |||
| NM_138980.3 | NP_620446.1 | 14 | 6172 | |||
| NM_138982.3 | NP_620448.1 | 14 | 6145 | |||
| XM_005263131.2 | XP_005263188.1 | 14 | 6039 | |||
| GenBank mRNA | AF052141.1 | - | 1 | 711 | ||