Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EIF2AK2 SIRNA Assays ›
Gene Symbol:
EIF2AK2
Gene Name:
eukaryotic translation initiation factor 2 alpha kinase 2
Gene Aliases:
EIF2AK1, PKR, PPP1R83, PRKR
Chromosome Location:
Chr.2: 37099210 - 37157065 on Build GRCh38
UniGene ID:
Hs.131431
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
n288894
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EIF2AK2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
XM_011532987.2
XP_011531289.1
17
9186
XM_017004503.1
XP_016859992.1
16
9053
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
Transcript Location
GenBankmRNA
BM930091.1
-
1
20
Chr. 2 - 37099366 - 37100019 [-] on Build GRCh38
GenBankmRNA
BU507028.1
-
1
160
Chr. 2 - 37099270 - 37100160 [-] on Build GRCh38
GenBankmRNA
CN404394.1
-
1
191
Chr. 2 - 37099411 - 37100176 [-] on Build GRCh38
GenBankmRNA
CR603904.1
-
1
2136
Chr. 2 - 37099241 - 37102135 [-] on Build GRCh38
GenBankmRNA
BU677520.1
-
1
149
Chr. 2 - 37099421 - 37100146 [-] on Build GRCh38
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPKK activity
positive regulation of cytokine production
transcription, DNA-templated
translation
protein phosphorylation
negative regulation of cell proliferation
response to mechanical stimulus
response to virus
response to toxic substance
regulation of translational initiation by eIF2 alpha phosphorylation
negative regulation of translation
peptidyl-tyrosine phosphorylation
evasion or tolerance by virus of host immune response
endoplasmic reticulum unfolded protein response
response to lipopolysaccharide
positive regulation of chemokine production
positive regulation of stress-activated MAPK cascade
response to vitamin E
negative regulation of osteoblast proliferation
regulation of protein phosphatase type 2A activity
cellular response to amino acid starvation
response to interferon-alpha
positive regulation of apoptotic process
negative regulation of apoptotic process
response to exogenous dsRNA
negative regulation of viral genome replication
innate immune response
protein autophosphorylation
positive regulation of NF-kappaB transcription factor activity
defense response to virus
regulation of NLRP3 inflammasome complex assembly
positive regulation of NIK/NF-kappaB signaling
regulation of hematopoietic progenitor cell differentiation
regulation of hematopoietic stem cell proliferation
regulation of hematopoietic stem cell differentiation
double-stranded RNA binding
protein kinase activity
protein serine/threonine kinase activity
eukaryotic translation initiation factor 2alpha kinase activity
non-membrane spanning protein tyrosine kinase activity
protein binding
ATP binding
protein phosphatase type 2A regulator activity
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-72rkq:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline