Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NDUFC1 SIRNA Assays ›
          Gene Symbol:
          NDUFC1
          Gene Name:
          NADH:ubiquinone oxidoreductase subunit C1
          Gene Aliases:
          KFYI
          Chromosome Location:
          Chr.4: 139289917 - 139302551 on Build GRCh38
          UniGene ID:
          Hs.84549
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n289324

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NDUFC1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001184986.1 NP_001171915.1 1 401
          NM_001184991.1 NP_001171920.1 1 401
          GenBank mRNA AK023115.1 - 1 1662
          CX163301.1 - 1 401
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA CN360331.1 - 1 405 Chr. 4 - 139300602 - 139301213 [-] on Build GRCh38
          GenBankmRNA BM771098.1 - 1 364 Chr. 4 - 139300602 - 139301173 [-] on Build GRCh38
          GenBankmRNA BM785365.1 - 1 364 Chr. 4 - 139300610 - 139301173 [-] on Build GRCh38
          GenBankmRNA BG676936.1 - 1 519 Chr. 4 - 139300661 - 139301313 [-] on Build GRCh38
          GenBankmRNA BM784018.1 - 1 364 Chr. 4 - 139295863 - 139301173 [-] on Build GRCh38
          GenBankmRNA CN360329.1 - 1 393 Chr. 4 - 139295926 - 139301202 [-] on Build GRCh38
          GenBankmRNA BI822563.1 - 1 246 Chr. 4 - 139295049 - 139301054 [-] on Build GRCh38
          GenBankmRNA BM747103.1 - 1 364 Chr. 4 - 139300648 - 139301173 [-] on Build GRCh38
          GenBankmRNA BX477751.1 - 1 495 Chr. 4 - 139300702 - 139301303 [-] on Build GRCh38
          GenBankmRNA BM745489.1 - 1 364 Chr. 4 - 139295942 - 139301173 [-] on Build GRCh38
          GenBankmRNA DA895687.1 - 1 527 Chr. 4 - 139300785 - 139301336 [-] on Build GRCh38
          GenBankmRNA BM756332.1 - 1 364 Chr. 4 - 139300602 - 139301173 [-] on Build GRCh38
          GenBankmRNA BU600840.1 - 1 213 Chr. 4 - 139295049 - 139301020 [-] on Build GRCh38
          GenBankmRNA BM768085.1 - 1 364 Chr. 4 - 139300602 - 139301173 [-] on Build GRCh38
          GenBankmRNA BI488544.1 - 1 587 Chr. 4 - 139300621 - 139301395 [-] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          oxidoreductase

          Gene Ontology Categories:

          Function(s) Process(es)

          mitochondrial electron transport, NADH to ubiquinone
          mitochondrial respiratory chain complex I assembly
          NADH dehydrogenase (ubiquinone) activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline