Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ERCC2 SIRNA Assays ›
          Gene Symbol:
          ERCC2
          Gene Name:
          ERCC excision repair 2, TFIIH core complex helicase subunit
          Gene Aliases:
          COFS2, EM9, TFIIH, TTD, TTD1, XPD
          Chromosome Location:
          Chr.19: 45349837 - 45370647 on Build GRCh38
          UniGene ID:
          Hs.487294
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n292398

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ERCC2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_011526611.2 XP_011524913.1 22 3619
          XM_017026467.1 XP_016881956.1 22 3533
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA AL135296.1 - 1 246 Chr. 19 - 45349939 - 45350567 [-] on Build GRCh38
          GenBankmRNA AL121529.1 - 1 246 Chr. 19 - 45350030 - 45350567 [-] on Build GRCh38
          GenBankmRNA BQ219568.1 - 464 Chr. - - [-] on Build
          GenBankmRNA DB636730.1 - 1 195 Chr. 19 - 45349838 - 45350516 [-] on Build GRCh38
          GenBankmRNA CR613691.1 - 1 993 Chr. 19 - 45349870 - 45351314 [-] on Build GRCh38
          GenBankmRNA BG389193.1 - 1 580 Chr. 19 - 45350176 - 45350900 [-] on Build GRCh38
          GenBankmRNA DB545503.2 - 1 176 Chr. 19 - 45349772 - 45350498 [-] on Build GRCh38
          GenBankmRNA BM050436.1 - 461 Chr. - - [-] on Build
          GenBankmRNA BG119711.1 - 1 454 Chr. 19 - 45350063 - 45350770 [-] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA helicase

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleotide-excision repair, DNA duplex unwinding
          response to hypoxia
          in utero embryonic development
          transcription-coupled nucleotide-excision repair
          nucleotide-excision repair
          nucleotide-excision repair, preincision complex stabilization
          nucleotide-excision repair, preincision complex assembly
          nucleotide-excision repair, DNA incision, 3'-to lesion
          nucleotide-excision repair, DNA incision, 5'-to lesion
          transcription initiation from RNA polymerase I promoter
          transcription elongation from RNA polymerase I promoter
          termination of RNA polymerase I transcription
          transcription from RNA polymerase II promoter
          transcription initiation from RNA polymerase II promoter
          transcription elongation from RNA polymerase II promoter
          7-methylguanosine mRNA capping
          protein phosphorylation
          apoptotic process
          response to oxidative stress
          chromosome segregation
          aging
          cell proliferation
          UV protection
          post-embryonic development
          viral process
          spinal cord development
          extracellular matrix organization
          bone mineralization
          central nervous system myelin formation
          nucleotide-excision repair, DNA incision
          multicellular organism growth
          hair cell differentiation
          embryonic cleavage
          erythrocyte maturation
          positive regulation of DNA binding
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          hair follicle maturation
          hematopoietic stem cell differentiation
          global genome nucleotide-excision repair
          regulation of mitotic cell cycle phase transition
          DNA binding
          ATP-dependent DNA helicase activity
          protein kinase activity
          protein binding
          ATP binding
          protein C-terminus binding
          DNA-dependent ATPase activity
          RNA polymerase II carboxy-terminal domain kinase activity
          5'-3' DNA helicase activity
          metal ion binding
          protein N-terminus binding
          4 iron, 4 sulfur cluster binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0