Search
Search

PDE12| Gene Symbol: PDE12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001322177.1 | NP_001309106.1 | 2 | 4671 | ||
| NM_177966.6 | NP_808881.3 | 3 | 4498 | |||
| GenBank mRNA | U79276.1 | - | 1 | 633 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | EB386946.1 | - | 1 | 342 | Chr. 3 - 57562198 - 57563037 [+] on Build GRCh38 | |