Search
Search

CDX2| Gene Symbol: CDX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011534875.2 | XP_011533177.1 | 3 | 2975 | ||
| XM_011534878.2 | XP_011533180.1 | 3 | 3137 | |||
| XM_011534879.2 | XP_011533181.1 | 3 | 3102 | |||
| GenBank mRNA | AK000205.1 | - | 1 | 260 | ||