Search
Search

MLXIP| Gene Symbol: MLXIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014938.5 | NP_055753.3 | 17 | 7933 | ||
| XM_006719292.3 | XP_006719355.1 | 17 | 7920 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR601671.1 | - | 1 | 1990 | Chr. 12 - 122144870 - 122147284 [+] on Build GRCh38 | |