Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SEC31A SIRNA Assays ›
          Gene Symbol:
          SEC31A THAP9-AS1
          Gene Name:
          SEC31 homolog A, COPII coat complex component
          THAP9 antisense RNA 1
          Gene Aliases:
          ABP125, ABP130, HSPC275, HSPC334, SEC31L1
          -
          Chromosome Location:
          Chr.4: 82818509 - 82900571 on Build GRCh38
          Chromosome Location:
          Chr.4: 82893452 - 82900916 on Build GRCh38
          UniGene ID:
          Hs.370024
          Hs.728916
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID n298883

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SEC31A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001318120.1 NP_001305049.1 1 163
          GenBank mRNA AB020712.1 - 1 130
          DA142666.1 - 1 163
          Gene Symbol: THAP9-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_034075.1 1 508
          NR_034076.1 1 508
          NR_034077.1 1 508
          GenBank mRNA BC020725.1 - 1 139
          BC045722.1 - 1 100
          BI831787.1 - 1 517
          DA084883.1 - 1 508
          DA361777.1 - 1 132
          DA923162.1 - 1 161
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA833028.1 - 1 160 Chr. 4 - 82894668 - 82900566 [-] on Build GRCh38
          GenBankmRNA BU178413.1 - 1 68 Chr. 4 - 82894385 - 82900476 [-] on Build GRCh38
          GenBankmRNA BX339948.2 - 1 141 Chr. 4 - 82894370 - 82900549 [-] on Build GRCh38
          GenBankmRNA BE880640.1 - 1 96 Chr. 4 - 82894534 - 82900504 [-] on Build GRCh38
          GenBankmRNA DA925660.1 - 1 99 Chr. 4 - 82894740 - 82900506 [-] on Build GRCh38
          GenBankmRNA CB127731.1 - 1 99 Chr. 4 - 82894723 - 82900507 [-] on Build GRCh38
          GenBankmRNA DB476991.1 - 1 159 Chr. 4 - 82894795 - 82900566 [-] on Build GRCh38
          GenBankmRNA BU506989.1 - 1 66 Chr. 4 - 82894394 - 82900474 [-] on Build GRCh38
          GenBankmRNA BI762414.1 - 1 128 Chr. 4 - 82894631 - 82900536 [-] on Build GRCh38
          GenBankmRNA DA568200.1 - 1 139 Chr. 4 - 82894620 - 82900547 [-] on Build GRCh38
          GenBankmRNA DA055649.1 - 1 161 Chr. 4 - 82894754 - 82900566 [-] on Build GRCh38
          GenBankmRNA DA615125.1 - 1 125 Chr. 4 - 82894781 - 82900536 [-] on Build GRCh38
          GenBankmRNA DA912133.1 - 1 100 Chr. 4 - 82894681 - 82900506 [-] on Build GRCh38
          GenBankmRNA CN429666.1 - 1 81 Chr. 4 - 82894685 - 82900483 [-] on Build GRCh38
          GenBankmRNA BM478668.1 - 80 Chr. - - [-] on Build
          GenBankmRNA DA082618.1 - 1 160 Chr. 4 - 82894739 - 82900566 [-] on Build GRCh38
          GenBankmRNA BM472827.1 - 161 Chr. - - [-] on Build
          GenBankmRNA DB472840.1 - 1 142 Chr. 4 - 82895694 - 82900550 [-] on Build GRCh38
          GenBankmRNA AL514590.3 - 1 50 Chr. 4 - 82894205 - 82900459 [-] on Build GRCh38
          GenBankmRNA DB030130.1 - 1 88 Chr. 4 - 82894751 - 82900496 [-] on Build GRCh38
          GenBankmRNA DA958580.1 - 1 101 Chr. 4 - 82894765 - 82900506 [-] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          vesicle coat protein

          Gene Ontology Categories:

          Function(s) Process(es)

          antigen processing and presentation of peptide antigen via MHC class I
          ER to Golgi vesicle-mediated transport
          protein transport
          antigen processing and presentation of exogenous peptide antigen via MHC class II
          IRE1-mediated unfolded protein response
          COPII vesicle coating
          response to calcium ion
          protein binding
          calcium-dependent protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0