Search
Search

RIF1| Gene Symbol: RIF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177663.1 | NP_001171134.1 | 35 | 8947 | ||
| NM_001177664.1 | NP_001171135.1 | 35 | 8850 | |||
| NM_001177665.1 | NP_001171136.1 | 34 | 9120 | |||
| NM_018151.4 | NP_060621.3 | 36 | 9025 | |||
| XM_005246665.3 | XP_005246722.1 | 36 | 8905 | |||
| XM_011511395.2 | XP_011509697.1 | 35 | 6161 | |||
| XM_017004422.1 | XP_016859911.1 | 36 | 8974 | |||
| XM_017004423.1 | XP_016859912.1 | 35 | 8812 | |||
| XM_017004424.1 | XP_016859913.1 | 35 | 8842 | |||
| XM_017004425.1 | XP_016859914.1 | 34 | 9150 | |||
| GenBank mRNA | BC072675.1 | - | 1 | 391 | ||