Search
Search

WNK1| Gene Symbol: WNK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011521005.2 | XP_011519307.1 | 1 | 474 | ||
| GenBank mRNA | AY231477.1 | - | 1 | 52 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA632847.1 | - | 1 | 70 | Chr. 12 - 851424 - 859375 [+] on Build GRCh38 | |