Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PIK3CA SIRNA Assays ›
Gene Symbol:
PIK3CA
Gene Name:
phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit alpha
Gene Aliases:
CLOVE, CWS5, MCAP, MCM, MCMTC, PI3K, PI3K-alpha, p110-alpha
Chromosome Location:
Chr.3: 179148114 - 179240093 on Build GRCh38
UniGene ID:
Hs.553498
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
n317862
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PIK3CA
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_006218.3
NP_006209.2
21
5158
GenBank mRNA
AK021510.1
-
1
369
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
Transcript Location
GenBankmRNA
CF136991.1
-
1
51
Chr. 3 - 179236115 - 179236669 [+] on Build GRCh38
GenBankmRNA
BX499689.1
-
1
106
Chr. 3 - 179236053 - 179236790 [+] on Build GRCh38
GenBankmRNA
BX115514.1
-
1
47
Chr. 3 - 179236112 - 179236748 [+] on Build GRCh38
Back To Top
More Information
Panther Classification:
Molecular Function -
kinase
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
liver development
vasculature development
glucose metabolic process
protein phosphorylation
phosphatidylinositol biosynthetic process
epidermal growth factor receptor signaling pathway
regulation of gene expression
regulation of phosphatidylinositol 3-kinase signaling
platelet activation
T cell costimulation
activation of protein kinase activity
positive regulation of peptidyl-serine phosphorylation
phosphatidylinositol-3-phosphate biosynthetic process
insulin receptor signaling pathway via phosphatidylinositol 3-kinase
Fc-epsilon receptor signaling pathway
Fc-gamma receptor signaling pathway involved in phagocytosis
ERBB2 signaling pathway
regulation of multicellular organism growth
regulation of cellular respiration
protein kinase B signaling
negative regulation of neuron apoptotic process
endothelial cell migration
hypomethylation of CpG island
phosphatidylinositol phosphorylation
vascular endothelial growth factor receptor signaling pathway
phosphatidylinositol-mediated signaling
T cell receptor signaling pathway
leukocyte migration
cardiac muscle contraction
adipose tissue development
cellular response to glucose stimulus
energy homeostasis
mitophagy in response to mitochondrial depolarization
negative regulation of fibroblast apoptotic process
regulation of genetic imprinting
negative regulation of anoikis
protein serine/threonine kinase activity
protein binding
ATP binding
kinase activity
1-phosphatidylinositol-3-kinase activity
protein kinase activator activity
phosphatidylinositol 3-kinase activity
1-phosphatidylinositol-4-phosphate 3-kinase activity
insulin receptor substrate binding
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0